accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ONN

CAS RN 99662-34-7

IUPAC Name: 4-(1H-pyrazol-1-yl)benzaldehyde
Synonyms: 4-(pyrazol-1-yl)benzaldehyde
Molecular Weight (g/mol): 172.19
Molecular Formula: C10H8N2O
InChi Key: PPGRDLZPSDHBIC-UHFFFAOYSA-N
SMILES: O=CC1=CC=C(C=C1)N1C=CC=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results