CAS RN 999-78-0
IUPAC Name:
4,4-dimethylpent-2-yne
Synonyms:
4,4-dimethylpent-2-yne
Molecular Weight (g/mol):
96.17
Molecular Formula:
C7H12
InChi Key:
FOALCTWKQSWRST-UHFFFAOYSA-N
SMILES:
CC#CC(C)(C)C
1
–
1
of
1
results
1
4,4-Dimethyl-2-pentyne, 97+%
CAS: 999-78-0 Molecular Formula: C7H12 Molecular Weight (g/mol): 96.17 MDL Number: MFCD00041613 InChI Key: FOALCTWKQSWRST-UHFFFAOYSA-N Synonym: 4,4-dimethyl-2-pentyne,2-pentyne, 4,4-dimethyl,acmc-20amsk,t-butylmethylacetylene,propyne, 1-tert-butyl,2,2-dimethyl-3-pentyne,4,4-dimethyl-pent-2-yne,tert-c4h9 c.$.cch3,tert-butylmethylacetylene,4,4-dimethyl-2-pentyne, PubChem CID: 136786 IUPAC Name: 4,4-dimethylpent-2-yne SMILES: CC#CC(C)(C)C