missing translation for 'onlineSavingsMsg'
Learn More

4,4-Dimethyl-2-pentyne, 97+%

Product Code. 11328305
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11328305 5 g 5g
11318305 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11328305 Supplier Thermo Scientific Alfa Aesar Supplier No. L10210.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 999-78-0 | C7H12 | 96.17 g/mol

4,4-Dimethyl-2-pentyne is used in organometallic reactions for regioselectivity of its insertion into the Pd-C bond of cyclopalladated complexes. It is also used to prepare pyrimidine-dione by reacting with isocyanate in the presence of nickel/ N-heterocyclic carbene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4-Dimethyl-2-pentyne is used in organometallic reactions for regioselectivity of its insertion into the Pd-C bond of cyclopalladated complexes. It is also used to prepare pyrimidine-dione by reacting with isocyanate in the presence of nickel/ N-heterocyclic carbene.

Solubility
Immiscible with water.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 999-78-0
Molecular Formula C7H12
Molecular Weight (g/mol) 96.17
MDL Number MFCD00041613
InChI Key FOALCTWKQSWRST-UHFFFAOYSA-N
Synonym 4,4-dimethyl-2-pentyne, 2-pentyne, 4,4-dimethyl, acmc-20amsk, t-butylmethylacetylene, propyne, 1-tert-butyl, 2,2-dimethyl-3-pentyne, 4,4-dimethyl-pent-2-yne, tert-c4h9 c.$.cch3, tert-butylmethylacetylene, 4,4-dimethyl-2-pentyne,
PubChem CID 136786
IUPAC Name 4,4-dimethylpent-2-yne
SMILES CC#CC(C)(C)C

Specifications

Density 0.718
Boiling Point 82°C
Flash Point −10°C (14°F)
Odor Petrol-like
Linear Formula (CH3)3CC≡CCH3
Refractive Index 1.4085
Quantity 5 g
UN Number UN3295
Beilstein 1733540
Solubility Information Immiscible with water.
Formula Weight 96.17
Percent Purity ≥97%
Chemical Name or Material 4,4-Dimethyl-2-pentyne
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.