accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CSiCH3H3CNHSiCH3CH3H3C

CAS RN 999-97-3

IUPAC Name: bis(trimethylsilyl)amine
Synonyms: hexamethyldisilizane
Molecular Weight (g/mol): 161.4
Molecular Formula: C6H19NSi2
InChi Key: FFUAGWLWBBFQJT-UHFFFAOYSA-N
SMILES: C[Si](C)(C)N[Si](C)(C)C
Percent Purity 
  • (4)
  • (5)
  • (3)
  • (6)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
Molecular Weight (g/mol) 
  • (3)
  • (7)
  • (6)
  • (1)
Boiling Point 
  • (6)
  • (5)
Physical Form 
  • (10)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (6)
  • (8)
  • (3)
  • (4)
  • (5)
  • (3)
  • (6)
  • (2)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (3)
  • (7)
  • (6)
  • (1)
  • (6)
  • (5)
  • (10)
  • (5)
  • (1)