Sustainability Consumables
Gibco™ BenchStable™ Cell Culture Media: Go Green by Reducing Laboratory Cold Storage Needs
April 2023 | 5 min read | By: Gibco
Gibco™ BenchStable™ Cell Culture Media: Go Green by Reducing Laboratory Cold Storage Needs
Advances in science and technology are critical to the continued evolution of society, including medical, environmental, and industrial breakthroughs. However, scientific research and development have also led to an increase in pollution and global warming. According to National Oceanic and Atmospheric Administration (NOAA) scientists, the year 2023 is likely to rank among the ten warmest years on record, with high temperatures and other adverse changes to climate likely due to carbon dioxide and other greenhouse gas emissions.1,2 Cold storage is a known substantial contributor to greenhouse gas emissions, with approximately 25% of laboratory energy consumption coming from cold storage.3
As an answer to this issue, and in accordance with their commitment to making the world healthier, cleaner, and safer, Thermo Fisher Scientific’s breakthrough Gibco™ BenchStable™ Media allows cell culture laboratories to store media at room temperature, reducing their cold storage footprint and decreasing negative environmental impacts. Based on calculations using energy consumption by Thermo Scientific™ TSX Series high-performance refrigerators, the global replacement of conventional media with BenchStable™ Media could result in a reduction in greenhouse gas emissions equivalent to removing over 5,000 passenger vehicles from the road each year.
Gibco™ BenchStable™ Media are available in the most commonly used formulations (DMEM, DMEM/F-12, RPMI 1640, and MEM) and have been proven to support cell culture performance at levels equivalent to standard cell culture media. BenchStable™ Media can be stored at room temperature prior to FBS supplementation for up to one year from the date of manufacture. This shelf life matches that of standard basal media under cold storage, with necessary vitamin and amino acid levels maintained over time in BenchStable™ media (Figure 1).
Furthermore, BenchStable™ Media bottles are provided in a recyclable, light-blocking paperboard box to protect media from the harmful effects of light exposure. Compared to the contributions of cold storage facilities on greenhouse gas emissions, this additional packaging has minimal environmental impacts.
24751_lab_reporter_figure_1.png
Figure 1. Components remain consistent in BenchStable™ Media. BenchStable™ RPMI (left) and BenchStable™ DMEM/F-12 (right) Media were maintained at room temperature (20 to 25°C) for 13 months. Samples taken at 1, 2, 3, 5, 7, 9, and 13 months were assessed for vitamin and amino acid concentrations via HPLC and compared with concentrations in lot-matched media stored at 2 to 8°C.
Comparable Growth and Functional Maintenance
The long-term effect of BenchStable™ Media on cell growth was assessed by comparing morphology and growth rates of cells cultured in 10% FBS-supplemented BenchStable™ medium to cells grown in a traditional medium. No significant performance or morphological differences were observed.
Results showed that doubling times for four cell types (adherent HeLa, adherent HEK293, adherent SH-SY5Y, and suspension Jurkat cells) were equivalent in BenchStable™ Media and traditional media types (Figure 2). SH-SY5Y, HEK293, and HeLa cells cultured in BenchStable™ Media were visually indistinguishable from those grown in standard media. Complex biological functions were also preserved in cells cultured in BenchStable™ Media, including the maintenance of expected gene expression patterns and healthy mitochondria (Figure 3).
24751_lab_reporter_figure_2.png
Figure 2. BenchStable™ Media support equivalent growth rates in long-term cultures. HeLa (MEM; pink), HEK293 (DMEM; blue), Jurkat (RPMI; green), and SH-SY5Y (DMEM/F-12; yellow) cells were grown in standard (closed points) and BenchStable™ (open points) Media supplemented with 10% FBS for 15 passages. Cell counts at each passage were used to calculate population doubling times.
24751_lab_reporter_figure_3.png
Figure 3. Cells cultured in BenchStable™ Media maintain healthy mitochondria. HeLa cells cultured in DMEM (left) and BenchStable™ DMEM (right) for eight passages were stained using tetramethylrhodamine methyl ester (TMRM) and Hoechst 33342 Solution. TMRM accumulates in active mitochondria, leading to a bright signal in healthy cells.
Equivalent Downstream Assay Performance
BenchStable™ Media were also shown to support cellular performance in downstream experiments and assays, including transfection and differentiation experiments. HeLa cells grown in BenchStable™ MEM exhibited transfection efficiencies equivalent to cells grown in standard MEM (Figure 4).
24751_lab_reporter_figure_4.png
Figure 4. Equivalent transfection efficiency in BenchStable™ Media cultures. HeLa cells cultured in MEM and BenchStable™ MEM for over five passages were transfected with GFP-containing plasmid DNA via Lipofectamine 3000. Transfection efficiency was measured in cells harvested 24 hours post-transfection by flow cytometry analysis, using untransfected cells as a negative control.
SH-SY5Y neuroblastoma cells cultured in standard and BenchStable™ DMEM/F-12 were assessed for their differentiation potential. Fluorescence microscopy data indicated that cells cultured in both media types exhibited good differentiation capabilities, confirming that BenchStable™ Media are compatible with complex and prolonged downstream assays (Figure 5).
24751_lab_reporter_figure_5.png
Figure 5. Effective differentiation in SH-SY5Y cells cultured in BenchStable™ Media. SH-SY5Y neuroblastoma cells cultured in DMEM/F-12 (left panels) and BenchStable™ DMEM/F-12 (right panels) were treated with 10 µM retinoic acid and allowed to differentiate for 5 days. Cells were stained with Alexa Fluor 488 phalloidin (green) and Hoechst 33342 Solution (blue) to allow for visualization of extended, branched neuron-like processes (top panels). Cholinergic neuron-like phenotypes (bottom panels) were shown by staining parallel cultures for tubulin (red) and choline acetyl transferase (ChAT, green).
An Eco-Friendly Cell Culture Solution
By enabling storage at room temperature prior to FBS supplementation and with performance equivalent to that of standard cell culture media, Gibco™ BenchStable™ media represent a greener option for cell culture laboratories.
References
1 Assessing the Global Climate in January 2023. www.ncei.noaa.gov/news/global-climate-202301
2 Annual 2019 Global Climate Report. National Centers for Environmental Information (National Oceanic and Atmospheric Administration). www.ncei.noaa.gov/access/monitoring/monthly-report/global/201913. Accessed: June 8, 2020.
3 Paradise A. Market Assessment of Energy Efficiency Opportunities in Laboratories. Center for Energy Efficient Laboratories. ET Project Number: ET14PGE7591, ET15SCE1070, ET14SDG1111. Issued March 12, 2015. www.etcc-ca.com/sites/default/files/reports/ceel_market_assessment_et14pge7591.pdf. Accessed June 8, 2020.