How Can You Optimise Electrochemistry Measurements for Improved Laboratory Efficiency?
March 2025 | 2 min read | By: Thermo Scientific
How Can You Optimise Electrochemistry Measurements for Improved Laboratory Efficiency?
Accuracy and reliability are fundamental requirements in any scientific or technical field, but they become critically important when measuring routine electrochemistry parameters, particularly in settings where key decisions are made based on the results.  Depending upon the laboratory setting, inconsistent or inaccurate data may not only impact upon your speed of analysis but may also potentially lead to costly errors, wasted resources or even public safety concerns.  
The challenges you face daily, from obtaining precise and reliable electrochemistry measurements, to the complexity of calibration, managing vast amounts of data, and ensuring efficient operations can be overwhelming and potentially lead to inaccuracies.
Now you can address these challenges with the newly launched Thermo Scientific™ Orion™ Pro Star Bench Meters – designed to help you simplify testing protocols and bring incremental improvements to your electrochemistry testing across a range of parameters such as; pH, DO, ORP, Ion and Conductivity measurements.   
25312_Lab_Reporter_image_2.jpg
Key Benefits
User-friendly operation:  Featuring an intuitive interface with a large 7-inch color touchscreen display operational with gloves, customizable layouts, guided navigation, and user-friendly troubleshooting assistance, all designed to enhance ease-of-use for technical and non-technical users alike
Comprehensive data management: With the ability to store up to 20 calibration logs per parameter and 10,000 time-stamped data log sets, you can maintain GLP compliant documentation with ease.  Easily export data to a printer, computer or USB flash drive for seamless data management and enhanced productivity
Practical design: Small footprint with vertical meter orientation and freestanding electrode arm to optimise bench space.  Constructed from IP-54 rated splash and dust-proof housing for dependable operation in harsh environments
These features combine to provide a user-friendly experience to help you reduce complexity in routine electrochemistry testing protocols.  Through implementing these improvements, you can uphold data accuracy and reliability whilst enhancing productivity.  Learn more about the range of Thermo Scientific Orion Pro Star Meters