New Innovative Technology Lab Equipment
RET basic Combined with IKA HUB for Smart Laboratory Workflows
September 2026 | 1 min read | By: IKA
RET basic Combined with IKA HUB for Smart Laboratory Workflows
IKA RET basic with IKA HUB
Safe remote control and monitoring – ideal for operation in a fume hood.
The RET basic combined with IKA HUB brings together the proven performance of the RET basic magnetic stirrer with the convenience of remote monitoring and control via the IKA HUB.
Designed for applications where safety is a priority, the system enables users to monitor and adjust process parameters without opening the fume hood. This minimizes exposure to hazardous substances while maintaining a safer working environment.
The included docking station allows the IKA HUB to be mounted directly outside the fume hood, allowing the hood to remain closed throughout the entire experiment while providing continuous access to critical data at all times.
Key benefits
  • Remotely monitor and control experiments for enhanced laboratory safety
  • Reduce exposure to toxic, volatile, or flammable substances
  • Ideal for high-temperature and long-duration applications
  • Simultaneously control and manage up to five connected devices
  • Improved workflow efficiency and experiment reproducibility
Experience smarter, safer laboratory control with the RET basic and IKA HUB.