Acetonitrile
Acetonitrile
Acetonitrile is the simplest organic nitrile and an aprotic solvent widely used for organic reactions and for manufacturing pharmaceuticals, pesticides, and perfumes. It is also used as a mobile phase in HPLC due to its low viscosity. Choose between various reagent grades depending on your needs.
Acetonitrile, HPLC for Gradient Analysis, Fisher Chemical™
HPLC for Gradient Analysis, meets analytical specification of Ph.Eur.
Acetonitrile, HPLC for Gradient Analysis, Fisher Chemical™
Acetonitrile, Advanced HPLC Grade for HPLC Gradient Analysis, Fisher Chemical™
High boiling point and low viscosity, perfect for your HPLC analysis.
Acetonitrile
Acetonitrile, HPLC Grade, 99.7+% min, Thermo Scientific Chemicals
Offers low noise in UV detection due to its low absorbance.
Acetonitrile
Complete Your Order - Great Deals
Thermo Scientific™ Hypersil™ BDS C18 Columns
BDS C18 Columns are used as robust, general-purpose columns for a wide range of applications in QC/QA labs.
Thermo Scientific™ Hypersil™ BDS C18 Columns
Ansell AlphaTec™ 02-100 Chemical Resistant, Five-Layer Laminated Gloves
Ansell AlphaTec™ 02-100 Laminated Gloves provide extreme resistance against a wide range of chemicals, including biological hazards.
Ansell AlphaTec™ 02-100 Chemical Resistant, Five-Layer Laminated Gloves
3M™ SecureFit™ Protective Eyewear with Anti-Fog Lens
3M™ SecureFit™ Protective Eyewear naturally adjusts to individual head sizes at the temples, for a customized fit to each person.
3M™ SecureFit™ Protective Eyewear with Anti-Fog Lens
Top Brands