Analytical Balances
Analytical balances are highly accurate and precise, and are designed to meet all your weighing needs within a chemical, industrial or pharmaceutical laboratory. They come with either an internal or external calibration system, with integrated software, and are offered with a wide range of weighing capacities.
Fisherbrand™ Analytical Balances
Featuring RS232 connectivity for easy communication, a backlit display, and a simple interface for uncomplicated operation, these balances are perfectly designed for your workplace.
Fisherbrand™ Analytical Balances
Sartorius™ Entris™ II Essential Analytical Balance
Entris™ II Essential balances ensure precise, fast, and chemically resistant weighing.
Sartorius™ Entris™ II Essential Analytical Balance
Ohaus™ Pioneer™ PX 224 Analytical Balance
The Pioneer PX delivers accurate, repeatable weighing for lab, industrial, and educational use.
Ohaus™ Pioneer™ PX 224 Analytical Balance
Selector Guide
Get the best-fit analytical balance for your lab protocols with our interactive selection guide.
Complete Your Order - Great Deals
Petri Dishes
Petri dishes for microbiology or cell culture come in various sizes and, materials, with reusable or disposable options.
Lorem Ipsum
Serological Pipettes
Serological pipettes transfer and mix liquids - they are available in glass or plastic, in sterile or non-sterile formats, and in various lengths.
Serological Pipettes
Exam Gloves
Examination gloves, made from latex, nitrile, PVC, or other polymers, protect hands in healthcare, labs and manufacturing, and come in various sizes, thicknesses, and colors.
Exam Gloves
FAQ
Analytical balances measure mass with extremely high precision, typically to 0.1 mg or better. They are commonly used in pharmaceutical, research, chemistry, and quality control laboratories where accurate sample preparation is essential.
Select a balance based on required readability, maximum capacity, calibration method, compliance requirements, and available workspace. The Sartorius, Fisherbrand and Ohaus brands offer models for routine and advanced applications.
Internal calibration provides convenient automated adjustment, while external calibration offers flexibility using certified weights. The best option depends on laboratory procedures and regulatory requirements.
Draft shields reduce airflow disturbances that can affect sensitive measurements. This helps improve weighing stability, repeatability, and overall measurement accuracy.
Analytical balances are widely used in pharmaceutical laboratories, academic research facilities, industrial quality control environments, chemical testing laboratories, and in life science applications.
Top Brands