Benchtop pH Meters
Benchtop pH Meters
Benchtop pH meters provide precise and repeatable pH measurement for research, quality control, environmental testing, food analysis and educational laboratories. This category includes advanced systems from the Mettler Toledo, Fisherbrand, Thermo Scientific and Cole Parmer brands, offering intuitive operation, reliable calibration workflows and compatibility with a wide range of pH electrodes. Whether performing routine measurements or demanding analytical applications, these benchtop pH meters deliver dependable results and efficient laboratory performance.
Mettler Toledo™ FiveEasy™ F20 pH/mV Meters
Compact, benchtop meter provides high quality pH/mV measurements with simple click of a button.
    Mettler Toledo™ FiveEasy™ F20 pH/mV Meters
    Fisherbrand™ accumet™ Basic AB315 Benchtop Laboratory pH/mV Meter
    Provides essential functions needed for common pH measurements without complicated functions, all at a budget-friendly price.
    Fisherbrand™ accumet™ Basic AB315 Benchtop Laboratory pH/mV Meter
    Mettler Toledo™ FiveEasy™ Plus FP20 pH/mV Meters
    Compact, benchtop meter provides high quality pH/mV measurements with simple click of a button.
      Mettler Toledo™ FiveEasy™ Plus FP20 pH/mV Meters
      Save Now - Exclusive Deals
      Complete Your Order - Great Deals
      Magnetic Stirrers
      Magnetic stirrers are lab tools that use a rotating magnetic field to spin a stir bar in a container, mixing liquids. They’re commonly used for tasks like preparing solutions, culturing cells, and carrying out chemical reactions.
      Magnetic Stirrers
      Electrochemistry Buffers
      Browse our buffer solutions for pH measurement, available as reference and calibration standards. Choose pH values from 1 to 10 in bottled or single-use pouches, offered in various colors and volumes.
      Electrochemistry Buffers
      Microcentrifuges
      Microcentrifuges are built for tubes under 5.0 mL. While benchtop centrifuges can use microtube rotors, microcentrifuges take up less space, helping keep the workspace less crowded.
      Microcentrifuges
      FAQ
      A benchtop pH meter offers higher measurement accuracy, stability, and feature integration compared to portable instruments.
      Typical laboratory-grade models achieve ±0.001 pH resolution and incorporate automatic temperature compensation (ATC), multi-point calibration, and data logging capabilities.
      They are designed for controlled environments such as analytical or QC labs, where precision, traceability, and compliance outweigh mobility.
      Handheld meters, while convenient, typically provide lower accuracy (±0.01 pH) and fewer calibration options, making benchtop units the standard for regulated or high-precision applications.
      pH readings are inherently temperature-dependent, since electrode potential varies with the Nernst equation slope (~59.16 mV/pH unit at 25 °C).
      Benchtop meters implement Automatic Temperature Compensation (ATC) by reading temperature via an integrated or external probe and adjusting the measurement accordingly.
      Without ATC, measurements can deviate significantly—especially in high-precision titrations or when comparing samples at different temperatures.
      Advanced meters allow manual input of sample temperature or real-time compensation curves, ensuring consistent results across varying thermal conditions.
      Accurate pH measurement depends on proper multi-point calibration using traceable buffer standards (commonly pH 4.01, 7.00, and 10.01).
      Benchtop instruments typically support 2–5 point calibration, automatically recognizing buffer values and applying slope and offset corrections.
      For regulated laboratories, calibration data should be logged, time-stamped, and linked to operator IDs to satisfy GLP/GMP or ISO 17025 documentation standards.
      Regular electrode maintenance (hydration, cleaning, and storage in proper solution) further ensures consistent response and extends electrode lifespan.
      Top Brands