Cell Function Reagents and Kits
Reagents and kits designed to visualize and examine cell function characteristics including dyes, stains, proliferation assays, viability reagents, and others.
FEATURED
Invitrogen™ LIVE/DEAD™ Viability/Cytotoxicity Kit, for Mammalian Cells
Two-color assay to determine cell viability. Useful with various fluorescence-detection methodologies.
Contec™ Sterile IPA
TRUSTED MTT REAGENT
Invitrogen™ CyQUANT™ MTT Cell Viability Assays
Complete package for a performing a cell-counting assay that can detect from 2000 to 250,000 cells.
Contec™ Sterile IPA
FOR 633 OR 635 NM EXCITATION
Invitrogen™ LIVE/DEAD™ Fixable Near-IR Dead Cell Stain Kit
Determine cell viability before fixation/permeabilization or elimination of biohazardous materials.
Contec™ Sterile IPA
Save Now - Exclusive Deals
Complete Your Order - Great Deals
FAQ
Cell function reagents and kits are used for:
  • Cell Viability and Proliferation Assays: Assessing cell health and growth (e.g., MTT, ATP assays)
  • Apoptosis and Necrosis Detection: Identifying cell death mechanisms (e.g., Annexin V/PI staining)
  • Cell Metabolism Studies: Measuring metabolic activities (e.g., Seahorse XF assays)
  • Cell Signaling Pathway Analysis: Investigating signaling pathways (e.g., phosphorylation assays)
  • Cytotoxicity Assays: Testing effects of compounds on cell viability (e.g., LDH release assays)
These procedures are crucial for studying cellular functions, disease mechanisms, and drug effects.
The key differences between cell function and cell structure reagents and kits are based on their applications and the aspects of cellular biology they target. Cell Function Reagents and Kits are designed to measure and analyze various cellular activities and processes such as cell viability, apoptosis, metabolism, signaling pathways, and cytotoxicity assays. Cell Structure Reagents and Kits focus on visualizing and studying the physical components and architecture of cells. Examples include staining of cellular organelles (nucleus, mitochondria), cytoskeleton components (actin, tubulin), and cell membrane markers. Cell Function is used for assessing dynamic cellular processes such as cell growth, death, metabolic activity, and response to stimuli. Cell Structure is used for imaging and analyzing the organization, morphology, and structural integrity of cells and their components.
When selecting Cell Function Reagents and Kits, consider the following:
  1. Ensure the reagents and kits are specific to your target cell function or marker. High sensitivity is crucial for detecting low-abundance targets.
  2. Verify that the reagents and kits are compatible with your cell type, experimental conditions, and detection methods (e.g., flow cytometry, fluorescence microscopy).
  3. Check if the reagents and kits have been validated for your specific application. Look for quality control data and performance benchmarks provided by the manufacturer.
  4. Assess the complexity of the protocols. Kits with straightforward, well-documented protocols can save time and reduce the potential for errors.
  5. Consider the cost-effectiveness of the reagents and kits, including any required ancillary materials. Also, ensure that they are readily available to avoid interruptions in your research.
Shop By Brand