24980_Merch_P1_Product
Cell Structure Reagents and Kits
Reagents and kits designed to visualise cell structure characteristics including dyes and stains for the examination of both cytoskeletal and internal cell components.
FEATURED
Invitrogen™ DAPI and Hoechst Nucleic Acid Stains
Popular cell-permeant nuclear counterstain that emits blue fluorescence when bound to dsDNA.
24980 Merch P2 Product 1
Invitrogen™ NucBlue™ Live ReadyProbes™ Reagent
Room-Temperature Stable
Ready-to-use Hoechst nuclear fluorescence stain provided in convenient dropper bottles.
Invitrogen™ Monomeric Cyanine Nucleic Acid Stains
Bright fluorescence signals, low background, and strong binding affinity with nucleic acid
Five distinct dyes for ultrasensitive fluorescence detection of nucleic acid in imaging and flow cytometry
24980 Merch P2 Product 3
Complete Your Order - Great Deals
FAQ
Cell structure reagents and kits are used for a variety of procedures, including:
  • Immunofluorescence (IF): Detecting and visualizing specific proteins or cell structures using fluorescently labeled antibodies
  • Western Blotting: Identifying and quantifying specific proteins from cell lysates
  • Flow Cytometry: Analyzing cell populations based on the expression of specific markers
  • Cell Staining: Highlighting and visualizing cell structures using dyes or stains
  • Immunohistochemistry (IHC): Detecting specific antigens in tissue sections using antibodies
These procedures help in studying cell morphology, protein localization, and cellular processes
The key differences between cell function and cell structure reagents and kits are based on their applications and the aspects of cellular biology they target. Cell Function Reagents and Kits are designed to measure and analyze various cellular activities and processes such as cell viability, apoptosis, metabolism, signaling pathways, and cytotoxicity assays. Cell Structure Reagents and Kits focus on visualizing and studying the physical components and architecture of cells. Examples include staining of cellular organelles (nucleus, mitochondria), cytoskeleton components (actin, tubulin), and cell membrane markers. Cell Function is used for assessing dynamic cellular processes such as cell growth, death, metabolic activity, and response to stimuli. Cell Structure is used for imaging and analyzing the organization, morphology, and structural integrity of cells and their components.
When selecting cell structure reagents and kits, consider:
  1. Specificity: Ensure reagents specifically target the desired cell structures or components
  2. Compatibility: Verify compatibility with your cell type and experimental conditions
  3. Sensitivity: Choose reagents with high sensitivity for accurate detection
  4. Ease of Use: Opt for kits with clear instructions and user-friendly protocols
  5. Quality & Reliability: Select products from reputable suppliers with consistent quality and reliability
Resources and Related Information
Agilent BioTek Imaging and Microscopy
Learn about Agilent BioTek automated cell imagers and microscopes, the unique Augmented Microscopy experience. Integration and automation of all steps from image capture to publication-ready data.
24980 Merch P8 Doc 1
Shop By Brand