Centrifuge Tubes
Find reliable Centrifuge Tubes from trusted brands like Eppendorf, Fisherbrand, Corning, Thermo Scientific, and Brand. Explore sterile options, conical and V-bottom formats, and lab-ready pack types designed for consistent performance in everyday spins and high-RCF protocols.
Falcon™ 50 mL High Clarity Conical Centrifuge Tubes
Ideal for cell centrifugation; pelleting; separation by density gradients.
Fisherbrand™ Easy Reader™ Conical Polypropylene Centrifuge Tubes
Imprinted clear black graduations ensure accurate readings.
Falcon™ 15 mL Conical Centrifuge Tubes
Excellent optical clarity and reliable performance.
Selector Guide
FAQ
Polypropylene Polystyrene tolerates higher RCF and broader temperatures; Polypropylene Polystyrene offers superior clarity for visual checks but lower RCF tolerance.
Use sterile, DNase/RNase-free tubes for cell culture, molecular work, or any contamination-sensitive protocol; non-sterile suits general sample prep.
It’s the highest centrifugal force the tube can safely withstand. Match your instrument’s max RCF with the tube’s rating to avoid deformation or leaks.
Top Brands