Chemical-Resistant Coveralls
Chemical Resistant Coveralls
One-piece garments manufactured from specialty chemical-resistant materials. Primarily designed to protect the wearer from liquid chemical splash or spray with options for providing additional protection against infective agents, radioactive contamination, and static electricity.
FEATURED
DuPont Tyvek 600 Plus White Hooded Coverall
Providing an ideal balance of protection, durability and comfort. 
DuPont™ Tyvek 600 Plus White Hooded Coverall
PROTECTION AND COMFORT
Fisherbrand Tyvek Cat III Coverall
Soft Polyethylene Non-woven Fabric
Offers more protection than equivalent SMS garments. 
Fisherbrand™ Tyvek™ Cat III Coverall
EXCELLENT BARRIER
DuPont Tyvek 500 Xpert Hooded Coverall
Robust Yet Lightweight
Composed of flash spun high density polyethlene. 
DuPont™ Tyvek 500 Xpert Hooded Coverall
Selector Guide
Save Now - Exclusive Deals
Complete Your Order - Great Deals
FAQ
A chemical-resistant coverall is expected to provide protection to the wearer from hazardous chemicals and other dangerous substances.
  • Resistance against penetration of chemicals
  • Mechanical resistance against abrasion, punctures or tears
  • In particular applications, antistatic properties and resistance against pathogens or radioactive material are required
  • The coverall should meet relevant safety standards and certifications (e.g., OSHA, ANSI, EN standards) specific to chemical protective clothing
Chemical protection suits are further classified according to the time it takes for a certain concentration of a chemical to penetrate through the protective suit. The longer the breakthrough time the higher the resistance to permeation and the more effective the chemical protection.
Protection Class
Breakthrough Times
1
> 10 minutes
2
> 30 minutes
3
> 60 minutes
4
> 120 minutes
5
> 240 minutes
6
> 480 minutes
Coveralls differ according to their ability to resist mechanical, chemical, and particulate elements.
Class I coveralls are intended for use in households and environments with low risk exposure.
Class III coveralls are designed for use in environments with high risk exposure.
The following types of Class III coverall are available:
Coverall Type
Protection Level
Type 1, 1a, 1b, 1c
Gas tight
Type 3
Protection against fluids
Type 4
Protection against spray
Type 5
Protection against particles
Type 6
Limited protection against spray
The requirements that need to be met for the various levels of chemical protection apparel are set in the following European standards:
EN 943-1: Type 1a, 1b and 1c
Protective clothing against liquid and gaseous chemicals, including liquid aerosols and solid particles – Part 1: Performance requirements for ventilated and non-ventilated ‘gas-tight’ (type 1)
EN 943-2: Type 1a ET and type 1b ET
Protective clothing against liquid and gaseous chemicals, including liquid (for emergency teams), aerosols and solid particles – Part 2: performance requirements for gas-tight (type 1) chemical protective suits for emergency teams (ET)
EN 14605: Type 3 and 4
Protective clothing against liquid chemicals – performance requirements for clothing with liquid-tight (type 3) or spray-tight (type 4) connections, including items providing protection to parts of the body only (types PB [3] and PB [4])
EN ISO 13 982-1: Type 5
Protective clothing for use against solid particulates – Part 1: performance requirements for chemical protective clothing providing protection to the full body against airborne solid particulates (type 5 clothing)
EN 13 034: Type 6
Protective clothing against liquid chemicals – performance requirements for chemical protective clothing offering limited protective performance against liquid chemicals (type 6 and type PB [6] equipment)
Furthermore, the following standard testing methods are prescribed for chemical protection apparel:
  • EN ISO 6529 Protective clothing – protection against chemicals – determination of resistance of protective clothing materials to permeation by liquids and gases
  • EN 1149-5 Protective clothing – electrostatic properties
  • EN ISO 16602 Protective clothing for protection against chemicals – classification, labelling and performance requirements
Consult your safety manager who needs to perform a risk assessment on the working environment. Share the specifics of your application. Based on these you will be able to determine together:
  • What work needs to be performed: The type and duration of the work will largely determine the protection level and class
  • What risks are associated with the work: Which chemicals are being used and which mixtures are being produced - are hazardous materials present in particulate, liquid or gaseous form? What is the temperature range of the working environment, does the application involve working with flammable or explosive substances, and is there increased risk of abrasion, tear or puncturing of the protective suit? Consider cumulative effects of the exposure to hazardous materials
  • What other protective equipment is required for the application and is this compatible with the chosen protective suit: Important considerations are the use of additional breathing apparatus, eye and face protection, hearing or head protection gear, gloves, protective boots, and securing belts etc
  • What options are available for decontamination of the protective gear: The above aspects will largely determine the use of disposable or reusable chemical-resistant coveralls and disposal or decontamination will need to be organized accordingly
  • Seek recommendations from your local Fisher Scientific Channel Account Manager or Safety Specialist, who will be able to advise on appropriate solutions
Resources and Related Information
Protective Solutions For Pharmaceutical Industry Applications
Find information on various Personal Protective Equipment including chemical resistant coveralls, suitable for different applications.
Protective Solutions For Pharmaceutical Industry Applications
Multi-Hazard Chemical Protection
A step-by-step guide to easily identify the right suit for your chemical task.
Multi Hazard Chemical Protection
Shop By Brand