24981_Merch_P1_Product
Chinese Hamster Ovary Cell Culture Media
Liquid solutions, reagents, and dry compounds are designed to support the growth, proliferation, and transfection of Chinese Hamster Ovary (CHO) cells. They are available in a range of formulations and volumes to suit your specific requirements.
FEATURED
Gibco™ ExpiFectamine™ CHO Transfection Kit
High-efficiency, cationic transfection reagent with optimized enhancer and culture feed.
25356 Merch P2 Product 1
Gibco™ ExpiCHO™ Expression Medium
Chemically-defined, animal origin-free medium
GMP-manufactured, serum-free, protein-free medium for high-density growth and transfection of CHO cells.
25356 Merch P2 Product 2
Greener Choice CHO Media
Reduce your environmental impact
Greener Choice products offer one or more environmental benefits over regular products.
25356 Merch P2 Product 2
Save Now - Exclusive Deals
Complete Your Order - Great Deals
Manual Single-Channel Pipettes
Manually operated single-channel pipettes for the transfer of measured amounts of liquid between different containers. Used with either universal or device-specific disposable pipette tips.
Universal Pipette Tips
Suitable for any genomic or biological application, these disposable tips are available in sterile and non-sterile versions for a variety of specific functions and in various styles and lengths.
Media Bottles
From iconic glass bottles to sturdy plastic ones, choose the right media bottles for your application from our diverse portfolio.
FAQ
Chinese Hamster Ovary (CHO) cells are typically cultured in several types of media, depending on the specific requirements of the experiment or production process. Commonly used media include:
  • Dulbecco's Modified Eagle Medium (DMEM): Often supplemented with 10% Fetal Bovine Serum (FBS) and other additives such as L-glutamine, non-essential amino acids, and antibiotics
  • Ham's F-12 Nutrient Mixture: Frequently used alone or in combination with DMEM (e.g., DMEM/F-12) and supplemented with FBS and other nutrients
  • RPMI-1640 Medium
  • Serum-Free Media: without the use of animal serum (e.g., CHO-S-SFM II, EX-CELL® CHO DHFR-, CD OptiCHO™ Medium)
  • Protein-Free Media: without the use of animal-derived proteins (e.g., CD CHO Medium, CHOgro® Expression Medium)
Chinese Hamster Ovary (CHO) cells are cultured in the following way:
  • Preparation: Use appropriate culture media (e.g., DMEM, Ham's F-12) supplemented with necessary nutrients
  • Seeding: Plate cells in a sterile culture vessel (e.g., flasks, plates) at the desired density
  • Incubation: Maintain cells in an incubator at 37°C with 5% CO₂
  • Feeding: Regularly replace the spent media with fresh media to provide nutrients and remove waste
  • Subculturing: Periodically pass the cells to new culture vessels to prevent over-confluence and maintain healthy growth
When selecting CHO cell culture media, consider:
  1. Nutritional Requirements: Ensure the media provides all necessary nutrients, including amino acids, vitamins, and minerals, to support optimal cell growth and productivity
  2. Serum Content: Decide between serum-containing, serum-free, or protein-free media based on the need for consistency, cost, and regulatory considerations. Serum-free and protein-free media are often preferred for biopharmaceutical production
  3. Cell Line Compatibility: Choose a media formulation that is compatible and optimized for the specific CHO cell line you are using
  4. Productivity and Quality: Evaluate how the media affects the yield and quality of the desired product, such as recombinant proteins or monoclonal antibodies
  5. Regulatory Compliance: For biopharmaceutical applications, ensure the media components meet regulatory standards and are free from animal-derived components if required
Resources and Related Information
Cell Culture Workflow From Culture to Discovery
Find a complete range of products for every step of your cell culture workflow, helping you to ensure success every time.
24981 Merch P8 Doc 1
3D Cell Culture Handbook – Culturing and Analysis of 3D Cell Models
An overview of the cell lines, culture conditions, and analysis techniques that are being used in the transition from 2D to 3D cell models.
24981 Merch P8 Doc 2
Cytiva Cell Culture Serum Products
Find information on a variety of serum products.
24981 Merch P8 Doc 3
Shop By Brand