24982_Merch_P1_Product
Classical and Basal Cell Culture Media
An extensive portfolio of high-quality classical and basal media formulations such as MEM, DMEM, and RPMI in convenient packaging configurations.
Gibco™ RPMI 1640 Medium (ATCC Modification)
Originally developed to culture human leukemic cells in suspension and as a monolayer.
Gibco™ RPMI 1640 Medium (ATCC modification)
Greener Choice Selection of Classical and Basal Cell Culture Media
Choose the Greener Solution and go with a Greener Choice product.
Greener Choice Selection of Classical and Basal Cell Culture Media
SureTRACE Classical and Basal Cell Culture Media
Get full traceability on your products and deliveries with SureTRACE marked items.
SureTRACE Classical and Basal Cell Culture Media
Save Now - Exclusive Deals
Complete Your Order - Great Deals
Fetal Bovine Sera
Discover a variety of animal-derived sera from leading brands, suitable for a variety of cell culture-related research and educational purposes.
Cell Culture Dishes
Cell culture dishes are disposable or reusable shallow containers specifically designed to support the growth and propagation of cells in culture. They are available in variety of sizes, in single or multi-well formats, and can be round or rectangular. They are typically made of borosilicate glass or clear plastic, allowing distortion-free microscopic observation.
Cell Culture Flasks
Cell culture flasks are essential tools for the growth of microbial, insect or mammalian cells. They are available in rectangular, U-shaped or angled forms, with different cap and neck styles. Single or multilayer flasks are available, with culture areas ranging up to 2500cm².
FAQ
Basal media are basic nutrient solutions that provide essential nutrients for cell growth. They are often used as a starting point for customizing cell culture conditions but are not sufficient for optimal cell growth by themselves and usually require supplementation. Examples include DMEM, RPMI-1640, and MEM (Minimum Essential Medium).
Complete media are basal media supplemented with additional components like fetal bovine serum (FBS), growth factors, and hormones to support optimal cell growth. An example is DMEM with 10% FBS. Complete media are ready-to-use solutions tailored for the optimal growth of specific cell lines or primary cells, providing all the nutrients and growth factors necessary for cell culture.
The different types of cell culture media include:
  • Basal Media: Basic nutrient solutions (e.g., DMEM, RPMI-1640)
  • Serum-Supplemented Media: Basal media with added serum (e.g., DMEM + 10% FBS)
  • Serum-Free Media: Formulated without serum, with specific growth factors (e.g., RPMI-1640 with specific supplements)
  • Chemically Defined Media: Composed of known quantities of all ingredients (e.g., Hybridoma-SFM)
  • Specialized Media: Tailored for specific cell types or applications (e.g., Neurobasal Medium)
  • Stem Cell Media: Designed for stem cell growth and maintenance (e.g., mTeSR1)
  • 3D Culture Media: For culturing cells in three-dimensional structures (e.g., Matrigel)
When selecting classical and basal cell culture media, consider the following:
  1. Cell Type Requirements: Ensure the media meet the specific nutritional and environmental needs of the cell type
  2. Growth Factors and Supplements: Determine if additional growth factors, hormones, or supplements are needed
  3. Serum Use: Decide whether to use serum-supplemented or serum-free media based on experimental needs
  4. Formulation Consistency: Choose media with consistent and reproducible formulations to ensure experimental reliability
  5. Compatibility: Ensure the media are compatible with the intended downstream applications and assays
Resources and Related Information
Cell Culture Media and Supplements Selector Guide
For antibody-derived molecule, vaccine, and viral vector production.
24982_Merch_P9_Doc1
Media Mastery for Cell Culture
Find information on everything Cell Culture Related.
24982_Merch_P8_Doc2
Shop By Brand