Controlled Environment Cleaners and Disinfectants
Cleaning and disinfectant solutions designed for specific use in controlled environments requiring minimal residue.
Contec™ Sterile IPA
Contec™ Sterile IPA is an alcohol blend of 0.2 micron filtered, filled and bagged in a Grade C (ISO Class 7) cleanroom.
Contec™ Sterile IPA
Contec™ NeutraKlean™ Detergent with Purified Water, Sterile
Ideal for the low level cleaning required on floors, walls, and benches after maintenance and to remove disinfectant residues and general soil.
Contec™ NeutraKlean™ Detergent with Purified Water, Sterile
Contec™ Non-Sterile CyChlor
Contains no quaternary ammonium or surfactant so it's very low residue, saving time on residue removal.
Contec™ Non-Sterile CyChlor
Save Now - Exclusive Deals
Complete Your Order - Great Deals
Controlled Environment Wipers
Experience exceptional cleanliness in cleanrooms and sterile environments with our high absorbance cleaning products. Designed to minimize shedding, they are perfect for general cleaning and spill control.
Controlled Environment Wipers
Cleanroom Gloves
Cleanroom gloves prevent particulate contamination and meet the stringent standards of pharmaceutical, medical device, and biotechnology manufacturers.
Cleanroom Gloves
General Purpose Wipes
General purpose wipers are designed for a variety of cleaning, disinfection, and absorption tasks in industrial and personal settings.
General Purpose Wipes
Wipers
Discover comprehensive information and buying guides for wipers, suitable for both general-purpose and controlled environments.
Wipers
Contamination Control and Controlled Environments
Everything cleanroom related.  Lean on our broad collection of contamination control products for efficiency and compliance.
Contamination Control and Controlled Environments
Disinfectants and Solutions for Critical Environments
Learn more about Contec’s cleanroom wipes, mops and disinfectants used in various industries across the globe including biotechnology, pharmaceutical, medical device, healthcare and other critical life science applications.
Disinfectants and Solutions for Critical Environments
Must Have
FAQ
  • Bleach (Sodium Hypochlorite): Commonly used in households and healthcare settings
  • Hydrogen Peroxide: Effective for disinfecting surfaces and wounds
  • Isopropyl Alcohol: Often used for sanitizing surfaces and skin
  • Quaternary Ammonium Compounds (Quats): Widely used in the healthcare and food service industries
  • Phenolic Compounds: Used in healthcare settings for disinfecting surfaces and equipment
In healthcare environments, quaternary ammonium compounds (Quats) are among the most commonly used disinfectants. They are favored for their broad-spectrum antimicrobial activity, effectiveness against a wide range of pathogens, and relatively low toxicity and corrosiveness compared to other disinfectants. Additionally, bleach (sodium hypochlorite) and alcohol-based disinfectants (such as isopropyl alcohol or ethanol) are also widely used for specific applications such as surface disinfection and hand sanitization.
To select the correct cleaners and disinfectants for controlled environments, consider the following:
  • Target Microorganisms: Identify the pathogens you need to eliminate
  • Surface Compatibility: Ensure the disinfectant is safe for your surfaces
  • Regulatory Compliance: Verify adherence to relevant standards (e.g., EPA, FDA)
  • Efficacy: Choose products with proven effectiveness and appropriate contact time
  • Safety: Select non-toxic, non-flammable options
  • Environmental Impact: Consider biodegradability and disposal requirements
  • Ease of Use: Assess practicality in preparation, application, and contact time
  • Residue: Determine if residues need rinsing off
  • Cost: Evaluate cost-effectiveness for your needs
  • Supplier Support: Ensure adequate support and documentation from the supplier
Top Brands