Controlled Environments Mops and Accessories
Designed specifically for controlled environments, our top-quality mops and accessories ensure cleanliness, safety, and compliance in critical spaces such as laboratories, cleanrooms, and pharmaceutical facilities. Choose from a range of durable, easy-to-clean mops, microfiber pads, and ergonomic handles to maintain optimal hygiene standards.
Contec™ Quiltec™ I Mop Head
Manufactured from 100% 2-ply polyester around a rayon core to generate very small numbers of particles and fibres. 
Contec™ Quiltec™ I Mop Head
SureTRACE Mops for Your Cleanroom
Sign up for the SureTRACE program now and start saving. 
SureTRACE Mops for Your Cleanroom
Greener Choice
Browse our sustainable options designed to help reduce the environmental impact of your laboratory operations.
Greener Choice
Useful Links
Save Now - Exclusive Deals
Complete Your Order - Great Deals
Contec™ Amplitude™ Sigma™ Cleanroom Wipes
Use these wipes for general cleaning and spill pickup in ISO Class 6 or above cleanroom applications. Amplitude™ Sigma™ Cleanroom Wipes are highly sorbent, have excellent durability and cleanliness, and are useful in a variety of settings.
Contec™ Amplitude™ Sigma™ Cleanroom Wipes
DuPont™ Tyvek 600 Plus White Hooded Coverall
Providing an ideal balance of protection, durability and comfort. DuPont™ Tyvek™ Taped Seam Hooded Coveralls are composed of flash spun high density polyethylene.
    DuPont™ Tyvek 600 Plus White Hooded Coverall
    Ansell BioClean™ Nerva BNAL Extra Long Nitrile Cleanroom Gloves
    Nitrile 16 in. elbow length gloves are resistant to a range of chemicals and reduce the risk of cross contamination.
      Ansell BioClean™ Nerva BNAL Extra Long Nitrile Cleanroom Gloves
      FAQ
      The three main types of mop are:
      Flat Mop: Features a flat head with a disposable or reusable pad that's often made of microfiber. Ideal for everyday cleaning and works well on flat surfaces.
      String Mop: Also known as a traditional mop, it has a bundle of absorbent strings or yarns. It's effective for deep cleaning and soaking up spills but can be harder to wring out and dry.
      Sponge Mop: Has a rectangular or cylindrical sponge head that is often equipped with a wringer mechanism. Suitable for cleaning tile and linoleum floors, it can absorb a considerable amount of liquid and is easy to wring out.
      In a controlled environment, such as a cleanroom or laboratory, a flat mop with a microfiber pad is generally the most effective. This type of mop is preferred for several reasons. Microfiber pads can trap and hold more dirt, dust, and microbes compared to traditional mops, which is crucial for maintaining strict cleanliness standards. They also tend to shed fewer particles than string or sponge mops, reducing the risk of contamination. Microfiber pads can also be easily removed and sterilized, ensuring that the mop remains hygienic between uses. Flat mops allow for more controlled application of cleaning solutions, which is important for maintaining the specific conditions required in controlled environments.
      • Opt for microfiber mop heads as they trap dust, dirt, and microbes effectively
      • Ensure the mop material is low-linting to minimize particle shedding
      • Flat mops are preferred for their ease of use and coverage. Choose disposable mop heads for single-use or reusable ones that can be sterilized, based on cleanroom protocol
      • Ensure mop compatibility with cleaning solutions. A lightweight, ergonomic handle reduces fatigue and improves efficiency
      • Ensure the mop and components are certified for cleanroom use
      Resources and Related Information
      Wipers
      Discover comprehensive information and buying guides for wipers, suitable for both general-purpose and controlled environments.
      Wipers
      Contec™ Mopping Systems for Critical Environments
      Whether cleaning or disinfecting walls, floors, ceilings or inside RABS, isolators and bio-safety cabinets, Contec has a mop or cleaning tool that meets the demanding requirements of today’s highly regulated pharmaceutical and healthcare industry. 
      Contec™ Mopping Systems for Critical Environments
      Biopharma Application
      From drug discovery to final filling and packaging, Micronova products remove bioburden and particles to foster cleanliness in research and production areas.
      Biopharma Application
      Shop By Brand