Controlled Environment Wipes
Controlled Environment Wipes
Experience exceptional cleanliness in cleanrooms and sterile environments with our high absorbance cleaning products. Designed to minimize shedding, they are perfect for general cleaning and spill control. Our extensive product range includes wipes, sponges, and wearable mitts, available in both disposable and reusable formats. Elevate your cleaning regimen with our reliable solutions today.
Contec™ Amplitude™ Sigma™ Cleanroom Wipes
Use these wipes for all ultra-critical applications and general purpose cleaning. 
Contec™ Amplitude™ Sigma™ Cleanroom Wipes
Micronova™ Irradiated Cleanroom Wipes
A range of low-linting, absorbent wipes that are ideal for cleaning, sanitizing, polishing, and drying in the cleanroom
Micronova™ Irradiated Cleanroom Wipes
Contec™ Polynit™ Cleanroom Wipes
Low in particles and extractables and with good sorbency for solvents.
Contec™ Polynit™ Cleanroom Wipes
Useful Links
Save Now - Exclusive Deals
Complete Your Order - Great Deals
Contec™ Sterile IPA
Provided sterile by gamma irradiated using a validated process at no less than 25 kGy. Contec™ Sterile IPA is an alcohol blend of 0.2 micron filtered, filled and bagged in a Grade C (ISO Class 7) cleanroom.
Contec™ Sterile IPA
Texwipe™ Alpha™ Polyester Series Swabs
Knitted and cleanroom laundered polyester head provides a neutral background for obtained sample because of its high cleanliness levels. Texwipe™ Alpha™ Swabs are widely recommended for surface sampling and cleaning validation work.
Texwipe™ Alpha™ Polyester Series Swabs
PURUS™ 30-Layer EnviroTack™ Contamination Control Mats
Exclusively from PURUS™, EnviroTack™ mats are the first ever contamination control products that contain GMO free, plant-based renewable materials, providing an environmentally friendly alternative to removing dirt and dust from foot traffic and equipment wheels.
PURUS™ 30-Layer EnviroTack™ Contamination Control Mats
FAQ
A cleanroom maintains low levels of airborne particles, while a controlled environment manages factors like temperature, humidity, and lighting without specific particle standards. Cleanroom wipes are used in industries requiring strict contamination control, such as pharmaceuticals, biotechnology and semiconductors, and undergo rigorous testing for particle generation, extractables, and fibers. Controlled environment wipes are suitable for less stringent environments where some level of contamination control is still necessary, such as in laboratories, medical device manufacturing, or electronics assembly.
Disinfectant wipes are generally not considered environmentally friendly for several reasons. They are made of non-biodegradable materials, including synthetic fibers like polyester or polypropylene, which do not break down easily in the environment. The disinfectants used can contain chemicals that may be harmful to aquatic life and ecosystems if not disposed of properly. Disinfectant wipes are also typically single-use, contributing to increased waste. However, there are some eco-friendlier options available, such as wipes made from biodegradable materials and those using environmentally safe disinfectants. It's important to check product labels and opt for these alternatives when possible.
Eco-friendly wipes minimize environmental impact by featuring:
  • Biodegradable Materials: Made from natural fibers like bamboo, cotton, or wood pulp, which break down more easily in the environment
  • Compostability: Certified to decompose without harmful residues
  • Sustainable Sourcing: Made from renewable resources
  • Non-Toxic Ingredients: Use of natural, non-toxic, and environmentally safe cleaning agents
  • Minimal Packaging: Reduced or recyclable packaging to lower overall waste
Always check for certifications like ASTM D6400 or EN 13432 for compostability
Resources and Related Information
Wipers
Discover comprehensive information and buying guides for wipers, suitable for both general-purpose and controlled environments.
Exam Gloves Guide
Contec™ Wipes for Critical Environments
Contec offer many different wipe substrates, edge technologies, configurations, and processing options for both sterile and non-sterile, dry and presaturated, knitted, nonwoven wipes and microfibre wipes.
Contec™ Wipes for Critical Environments
Revolve - A Sustainable Solution For Your Cleanroom Environment
Learn more about the Revolve line of wipers, the first cleanroom products made from upcycled polyester.
Revolve - A Sustainable Solution For Your Cleanroom Environment
Shop By Brand