Electronic Multichannel Pipettes
Electronic Multichannel Pipettes are used with disposable pipette tips to simultaneously transfer multiple measured amounts of liquid between various format labware. Compatible with 96-well and 384-well microplates, they may have adjustable tip spacing or possess a programmable interface.
FEATURED
Thermo Scientific™ E1-ClipTip™ Bluetooth™ Electronic Multichannel Pipettes
Enjoy pipetting again with optimum functionality for every application and user preferences.
Contec™ Sterile IPA
EASY-TO-READ
Thermo Scientific™ Finnpipette™ Novus Multichannel Pipettes
Speed up repetitive dispensing into 96- and 384-well microplates.
Contec™ NeutraKlean™ Detergent with Purified Water, Sterile
READY-TO-USE
Sartorius Picus™ 2 Electronic 8-Channel Pipette
Reliable, ergonomic pipetting for all users, with customizable programs and safety features, as well as app connectivity.
Contec™ Non-Sterile CyChlor
Complete Your Order - Great Deals
Universal Pipette Tips
Universal pipette tips are compatible with pipettes from various manufacturers and have comparable performance to branded tips.
Universal Pipette Tips
Pipette Specific Tips
Check out our broad range of pipette tips for specific pipetting devices.
Pipette Specific Tips
FAQ
Electronic multichannel pipettes reduce the risk of repetitive strain injury and improve accuracy by automating pipetting steps. They also allow for programmable functions like multi-dispensing, mixing, and dilution, which save time and increase reproducibility in high-throughput workflows.
They are commonly available in 8-channel and 12-channel formats, designed to match standard 96-well microplates. Some advanced models can even adjust spacing to accommodate different plate or tube formats, making them more versatile across applications.
Most electronic multichannel pipettes are designed to work with universal pipette tips, but optimal performance is achieved with tips recommended by the manufacturer. Using the correct tips ensures airtight sealing, precise dispensing, and prevents sample contamination.
Shop By Brand