Exam Gloves
Examination gloves are made from a variety of materials including latex, nitrile, polyvinyl chloride, and other polymers. In addition to their use in clinical or healthcare environments, examination gloves are also worn to protect the hands in quality control laboratories, manufacturing plants, and R&D laboratories. Browse through our wide selection of exam gloves in different sizes, thicknesses, and colors.
Useful Links
Selector Guide
Use this interactive guide to find the perfect exam gloves for your application. Just select a feature, walk through the options, and our recommendations will be displayed right here.
Save Now - Exclusive Deals
Product Code 11819620
Product Code 10754175
Product Code 15289373
Product Code 15569471
Product Code 16885440
Product Code 17929980
Product Code 17844871
Product Code 16386901
Product Code 17849662
Product Code 17894861
Complete Your Order - Great Deals
Product Code 16594231
Product Code 11895843
Product Code 15807111
Product Code 12394739
Must Have
Product Code 30689993
Product Code 15299363
Product Code 17621036
Product Code 16499405
Product Code 17894881
Product Code 17899662
Product Code 13178646
Product Code 17879662
Product Code 30689996
Product Code 30689997
Product Code 30689992
Product Code 30689990
Product Code 30689995
FAQ
Nitrile gloves are better suited for medical examinations, procedures requiring a high level of protection, and handling chemicals due to their durability, chemical resistance, and comfort. Vinyl gloves are more appropriate for low-risk, non-hazardous tasks and situations where cost is a significant concern. Overall, nitrile gloves are generally considered superior for medical and high-risk environments, while vinyl gloves are suitable for low-risk tasks and situations where cost savings are important.
Disposable gloves are a broad category of single-use gloves used in various industries like food service, cleaning, and industrial work. They are made from materials such as latex, nitrile, vinyl, and polyethylene, and their standards vary by intended use. Exam gloves are a specific type of disposable glove designed for medical examinations and procedures, primarily used in healthcare settings to prevent cross-contamination. They must meet strict medical standards (e.g., FDA, ASTM) and are typically made from latex, nitrile, or vinyl. Exam gloves are available in both sterile and non-sterile versions and often come in dispenser boxes for easy access. In summary, while all exam gloves are disposable, not all disposable gloves are exam gloves.
No, they are not the same. Examination gloves are designed for medical examinations and procedures, providing a barrier to prevent cross-contamination. They are available in both sterile and non-sterile versions and must meet specific medical standards (e.g., FDA, ASTM). Clean gloves are generally used for tasks requiring cleanliness but not medical sterility, such as handling food or cleaning. They are usually non-sterile and do not need to meet stringent medical standards.