General Purpose Wipers
General Purpose Wipes
General purpose wipers are designed for a variety of cleaning, disinfection, and absorption tasks in industrial and personal settings. Manufactured from a variety of materials, with both disposable and reusable options, they are available as individual wipes, in bulk boxes, and in rolls.
Tork Heavy-Duty Cleaning Cloth W1/2/3
Low-lint, high absorbency and chemically inert wipes for delicate tasks and instrumentation in a range of applications.
Tork Heavy-Duty Cleaning Cloth W1/2/3
Tork Cleaning Cloth W1/2/3
For versatile cleaning and wiping in process and maintenance environments.
Tork Cleaning Cloth W1/2/3
Kimberly-Clark™ Kimtech™ Science Delicate Task Wipers 7558
Low-lint, high absorbency, and chemically inert precision wipes for delicate tasks and instrumentation in a range of applications..
Kimberly-Clark™ Kimtech™ Science Delicate Task Wipers 7558
Complete Your Order - Great Deals
Chemgene MedLab Multi-Surface Disinfectant Concentrate
CHEMGENE MEDLAB Multi-Surface Disinfectant Concentrate delivers a new, unique formulation designed for specialist laboratory settings. The combination of active substances in CHEMGENE ensure that equipment can be routinely cleaned and disinfected to a consistently high standard.
Chemgene MedLab Multi-Surface Disinfectant Concentrate
Exam Gloves
Our range of Exam Gloves are available in different sizes, thicknesses, and colors. Made from a variety of materials, including latex, nitrile and polyvinyl chloride, they are suitable for use in clinical/healthcare environments and in quality control laboratories, manufacturing plants, and R&D laboratories.
Exam Gloves
Tork Floor Stand Dispenser W1
The Tork floor stand dispenser in Performance design is developed for safety and stability with its reliable tear-off and easy-load features.
    Tork Floor Stand Dispenser W1
    FAQ
    General purpose wipes are versatile cloths or towelettes for everyday cleaning in homes, offices, and commercial settings. They handle tasks like wiping surfaces, cleaning spills, dusting, and sanitizing. Made from paper, cloth, or non-woven fabrics, they can be dry or pre-moistened. These wipes are valued for their convenience, ease of use, and effectiveness. Controlled environment wipes on the other hand, are designed for cleanrooms, labs, and areas requiring contamination control. They are made from low-lint, non-shedding materials such as polyester or microfiber and may be sterile or have low ionic contaminants. Produced under strict quality controls to minimize contamination, they meet rigorous standards like ISO.
    Both dry and wet wipes offer distinct advantages depending on the specific needs and preferences of the user. Dry wipes allow users to add their preferred type and amount of chemicals, disinfectants, or simply water, offering flexibility for different tasks while pre-moistened and ready to use wipes offer convenience for on the go cleaning and personal hygiene. Generally dry wipes are more economical than pre-moistened wipes, especially when purchased in bulk. Dry wipes can be stored for longer periods without the risk of drying out, unlike wet wipes. Wet wipes are available in various formulations, including antibacterial, disinfectant or specialty wipes for specific tasks.
    Industrial wipes are absorbent, durable cleaning materials designed for use in various industrial and commercial settings. They are typically used for tasks such as cleaning machinery, wiping down surfaces, removing grease and oil, and general maintenance. Industrial wipes come in a variety of materials, including cloth, paper, and synthetic fibers, and they may be treated with specific chemicals to enhance their cleaning capabilities or to meet specific industry standards. They are valued for their strength, absorbency, and versatility in handling tough cleaning jobs. 
    Resources and Related Information
    Wipers
    Discover comprehensive information and buying guides for wipers, suitable for both general-purpose and controlled environments.
    Wipers
    Tork Hygiene Solutions
    Achieve the highest service levels and improve efficiency with Tork Hygiene Solutions.
    Wipers
    Top Brands