25343_Merch_P1_Product
Goat Secondary Antibodies
Browse our extensive range of goat secondary antibodies targeting various species. Choose from monoclonal or polyclonal antibodies, available in conjugated, unconjugated, and other forms, suitable for a wide range of applications to meet your specific needs.
FEATURED
IgG (H+L) Cross-Adsorbed Goat anti-Mouse, Alexa Fluor™ 488, Invitrogen™
Polyclonal Goat Secondary Antibody, cross-adsorbed against human IgG and human serum.
25343_Merch_P2_Product1
IgG (H+L) Highly Cross-Adsorbed Goat anti-Mouse, Alexa Fluor™ Plus 488, Invitrogen™
Polyclonal Antibodies for Low Abundancy Targets
A brighter signal with lower cross-reactivity gives higher sensitivity and better signal-to-noise for your critical experiments.
25343_Merch_P2_Product2
Amersham™ CyDye™ 700 Goat-Anti-Mouse Secondary Antibody, Cytiva
Easy-to-Handle Near IR-Labeled Antibodies
Possible to multiplex more than one target protein on a single blot. Stability allows rescanning of membranes for future studies.
25343_Merch_P2_Product3
Save Now - Exclusive Deals
Complete Your Order - Great Deals
Thermo Scientific™ Blocker™ BSA (10X) in PBS
Block membranes, plates, and other samples for Western blot, ELISA, and other detection methods with these ready-to-use 10X solutions of bovine serum albumin (BSA) in PBS.
Streptavidin-Biotin Binding Products
Explore our range of Streptavidin–Biotin binding products, designed for the identification and purification of proteins and nucleic acids in assays such as ELISA, IHC, and Western blot.
FAQ
Goat secondary antibodies are antibodies produced in goats that bind specifically to primary antibodies from another species (e.g., rabbit, mouse). The goat's immune system recognizes these foreign IgGs and produces antibodies against them. The resulting antibodies are highly specific to the IgG of the species they were raised against. For example, Goat anti-rabbit: Binds to rabbit IgG. Goat anti-mouse: Binds to mouse IgG. Goat anti-human: Binds to human IgG.
They are used in various immunoassays (Western blotting, ELISA, immunohistochemistry, immunofluorescence) to detect and visualize target antigens. These antibodies are often conjugated to reporter molecules like enzymes (HRP), fluorophores (FITC), or biotin for detection.
The number of times you can use a secondary antibody depends on the antibody's stability, storage conditions, and the specific application. Typically, secondary antibodies can be used multiple times if they are stored properly usually at -20°C for long-term storage or 4°C for short-term, and if they have not been contaminated.  Using sterile techniques and aliquoting the antibody to avoid repeated freeze-thaw cycles can extend its usability. The number of uses can also depend on the type of experiment (e.g., Western blotting, immunohistochemistry) and the concentration required. For Western blotting, a secondary antibody can often be reused multiple times, sometimes up to 10-20 times, if stored correctly and used at the recommended dilution. Always refer to the manufacturer's guidelines for the most accurate information.
When selecting goat secondary antibodies, consider the following five factors:
  1. Species Specificity: Ensure the secondary antibody is specific to the primary antibody's host species (e.g., goat anti-rabbit for rabbit primary antibodies). 
  2. Conjugation: Choose the appropriate conjugate for your detection method, such as enzymes (HRP, AP), fluorophores (FITC, Alexa Fluor), or biotin.
  3. Affinity and Purity: Select antibodies with high affinity and purity to ensure specific binding and minimal background noise. 
  4. Application Compatibility: Ensure the secondary antibody is suitable for your specific application (e.g., Western blotting, ELISA, immunohistochemistry, immunofluorescence). 
  5. Validation and Reviews: Check for validation data and reviews to confirm the antibody's reliability and performance in similar experiments.
Resources and Related Information
ELISA Guide
A clear and easy guide to ELISAs.
Labels and Ribbons
Why Choose Bio-Techne Antibodies?
From benchtop research to clinical applications - be confident in your decision when selecting  Bio-Techne antibodies.
Labels and Ribbons
Shop By Brand