Combination Hotplate Stirrers
Hotplate stirrers are popular laboratory devices with which solutions or liquids can be heated and mixed simultaneously. Different models are available with specific functions for your individual needs: from good quality reliable analog hotplate stirrers to digital models with precise temperature and speed control, safety protection and other advanced features, such as timers and programmable functions.
Useful Links
Save Now - Exclusive Deals
Product Code 15673687
Product Code 15323528
Product Code 17961971
Product Code 15215617
Complete Your Order - Great Deals
Must Have
Product Code 15353538
Product Code 17814447
Product Code 17887398
Product Code 17745868
Product Code 15400484
FAQ
A hotplate stirrer combines two essential laboratory functions, precise heating and magnetic stirring, within one unit. While a hotplate alone provides uniform surface heating and a magnetic stirrer enables mixing via a rotating magnetic field, the integrated design of a hotplate stirrer allows controlled temperature and agitation simultaneously. This is critical for sample dissolution, reagent preparation, titrations, evaporation, and reaction kinetics studies. High-end models feature PID temperature control, variable stirring speed, and compatibility with external temperature probes (PT100 or RTD sensors) for direct feedback from the sample solution.
The top-plate composition directly impacts heating efficiency, chemical resistance, and durability:
Ceramic plates deliver fast heating, are chemically inert, and resist acid corrosion, ideal for volatile or corrosive samples.
Aluminum or anodized alloy plates provide superior heat distribution and are suited for heavy glassware or viscous solutions.
Infrared or microprocessor-controlled heating ensures uniform thermal gradients and faster ramp-up times. For temperature-critical reactions, advanced models integrate closed-loop PID control and external probe compensation, maintaining stability within ±1 °C of the set point.
Modern laboratory hotplate stirrers incorporate multiple safety and compliance features to meet regulatory and operational requirements:
Over-temperature protection and hot-surface warning indicators to prevent burns or sample degradation.
Automatic shut-off when a probe disconnects or the plate exceeds safety thresholds.
Encapsulated electronics and chemical-resistant housings to protect against spills and vapors.
Magnetic coupling detection that halts stirring if the stir bar decouples. Digital and network-capable models may also log calibration data, provide error diagnostics, and support GLP/GMP documentation, ensuring traceability and reproducibility in regulated labs.