Isopropanols
Isopropanols
Isopropanol (IPA), also known as 2-propanol, is a versatile solvent used across analytical chemistry, chromatography, cleaning, and sample preparation. Choosing the right grade matters: HPLC and LC/MS grades help reduce background noise and improve method reproducibility, while ACS and analytical grades support dependable routine workflows. In this selection, you’ll find isopropanols suited to chromatography, trace analysis, and everyday laboratory needs—available in popular pack sizes for efficient lab stocking. Explore trusted options from the Fisher Chemical™, Honeywell™ (including Fluka™ and Research Chemicals™ ranges), and JT Baker™ brands to match your application requirements, documentation needs, and purity expectations.
Isopropanol, for HPLC, Fisher Chemical™
Amber Glass Bottle, 2.5 L.
Isopropanol, for HPLC, Fisher Chemical™
Isopropanol, Optima™ LC/MS Grade, Fisher Chemical™
Glass Bottle, 2.5 L.
Isopropanol, 99.5+%, pure
Isopropanol, Certified AR for Analysis, Fisher Chemical™
2.5 L.
2-Propanol, CHROMASOLV™ LC-MS, Honeywell Riedel-de Haën™
Complete Your Order - Great Deals
Isopropanols for HPLC Performance
Reduce variability and support consistent chromatography with trusted HPLC-grade IPA options.
Isopropanols for HPLC performance
Ready for LC/MS Workflows
Choose LC/MS-suited IPA to help minimize background and protect sensitivity in trace-level analysis.
Ready for LC/MS workflows
FAQ
Isopropanol is used for chromatography, sample preparation, cleaning, molecular biology workflows, and analytical testing. High-purity grades from Fisher Chemicals, Thermo Scientific, and Fluka brands/portfolios help ensure reliable and reproducible laboratory results.
Select the grade based on application requirements. HPLC and LC/MS grades are ideal for chromatography, analytical grades support testing laboratories, while molecular biology grades are suitable for DNA, RNA and life science workflows.
Leading options include the Fisher Chemicals, Thermo Scientific, Fluka, Honeywell Riedel-de Haën and MP Biomedicals portfolios . These brands provide a range of purity levels and package sizes for research and industrial laboratories.
HPLC-grade isopropanol is commonly used in liquid chromatography systems, mobile phase preparation and sample analysis where low impurity levels are required to maintain instrument performance and analytical accuracy.
Higher-purity isopropanol reduces contamination risks, improves reproducibility and supports compliance in regulated environments. Choosing the appropriate grade helps protect instruments and ensures dependable experimental outcomes.
Top Brands