nginx
Life Science Buffers
Life Science Buffers
Life science buffers help maintain stable pH and ionic strength across workflows like cell culture, sample preparation, protein work, and molecular biology. From ready-to-use solutions (e.g., PBS and wash buffers) to concentrated and powder formats for flexible dilution, choosing the right buffer supports reproducibility and reduces variability between runs. Explore trusted options from Affymetrix, BD Difco, Corning Media Tech, Fisher Bioreagents, and Fisher Chemicals to match your application needs.
PBS Tablets, Phosphate Buffered Saline, Fisher BioReagents™
Phosphate Buffered Saline Tablets, CAS Number 7647-14-5, 7447-40-7, 100 tablets, Clear, Gray, White, 5/8 in., 600 SEC max., CAUTION!
    PBS Tablets, Phosphate Buffered Saline, Fisher BioReagents™
    Thermo Scientific™ Pierce™ Concentrated Buffer Stocks (10X and 20X)
    Our variety of Pierce Concentrated Buffer stock solutions are ready to use without having to weigh and dissolve dry ingredients or adjust the pH with concentrated acid or base. Simply dilute the liquid stock solution with pure water and proceed with your experiment.
      Thermo Scientific™ Pierce™ Concentrated Buffer Stocks (10X and 20X)
      PBS, Phosphate Buffered Saline, 10X Solution, Fisher BioReagents™
      Save time and simplify your buffer preparation step by using concentrated Fisher BioReagents 10X PBS solution - simply dilute as needed
        PBS, Phosphate Buffered Saline, 10X Solution, Fisher BioReagents™
        Complete Your Order - Great Deals
        Round Plastic Bottles
        Explore our extensive range of durable plastic bottles, designed for a multitude of applications. With their versatility and high-quality materials, they are perfect for storing chemicals, managing waste, and more. Find the ideal plastic bottle you've been searching for today.
        Round Plastic Bottles
        Magnetic Hotplate Stirrers
        Magnetic stirrers are laboratory tools used to stir liquids, usually in a container. They create a rotating magnetic field that causes a stir bar placed in the container to rotate, which in turn stirs the liquid. Magnetic stirrers are used for a variety of laboratory applications, such as mixing solutions, culturing cells, and performing chemical reactions.
        Magnetic Hotplate Stirrers
        FAQ
        Life science buffers stabilise pH and ionic strength to protect samples and improve reproducibility in cell culture, molecular biology, and protein workflows. Common examples include PBS, Tris, HEPES, and TE buffers. Choose the grade and format that match your assay sensitivity and handling needs.
        PBS and DPBS are both phosphate-buffered saline solutions used for washing and dilution. DPBS often includes specific salts suited to certain cell handling steps, while PBS is widely used for general rinsing and buffer preparation. Select based on your protocol requirements and cell type compatibility.
        HEPES is commonly used for strong pH buffering near physiological pH, especially when CO₂ control is limited. It helps reduce pH drift during handling outside incubators. Check concentration, sterility, and grade to match cell culture or molecular biology workflows.
        Concentrated buffers are diluted with high-quality water to reach 1X working strength, helping save storage space and reduce shipping. Mix thoroughly and verify pH if your protocol requires tight control. For sensitive workflows, consider molecular biology grade options from brands like Fisher Bioreagents or Affymetrix.
        Molecular biology grade buffers are typically manufactured to minimise contaminants like nucleases and proteases, supporting PCR, cloning, and RNA workflows. General-purpose grades may be suitable for routine washes and non-sensitive steps. Match the grade to downstream risk and required assay performance.
        Top Brands