Manual Single-Channel Pipettes
Manually operated single-channel pipettes are used to transfer a measured amount of liquid between different containers. Sample contamination is avoided through the use of disposable pipette tips (either universal or device specific).
FEATURED
Thermo Scientific™ Finnpipette™ F2 Variable Volume Pipettes
Meet the challenges of highly demanding applications with the sleek, rugged design of these fully autoclavable pipettes.
Contec™ Sterile IPA
EASY-TO-READ
Eppendorf Research™ Plus Pipette
Ergonomic, lightweight, and designed for precision in every transfer.
Contec™ NeutraKlean™ Detergent with Purified Water, Sterile
READY-TO-USE
Thermo Scientific™ Finnpipette™ F2
Built tough. Autoclavable. Precision you can rely on in every drop.
Contec™ Non-Sterile CyChlor
Complete Your Order - Great Deals
Universal Pipette Tips
Our range of Universal Pipette Tips are compatible with pipettes from various manufacturers and have comparable performance to branded tips.
Universal Pipette Tips
Pipette Specific Tips
Our range of Pipette Specific Tips are disposable and designed to fit specific manufacturers’ single and multichannel pipettes.
Pipette Specific Tips
FAQ
They are ideal for transferring precise liquid volumes in applications where accuracy and reliability are critical, such as preparing samples, running assays, or transferring reagents between tubes and plates.
Manual single-channel pipettes come in fixed or adjustable volume formats. Choose a pipette where your most frequently used volume falls in the middle of its range—for example, a 20–200 µL pipette is most accurate around 100 µL.
Regular calibration, proper storage in an upright position, and using high-quality compatible tips are essential. Avoid laying pipettes flat when loaded with liquid, as this can cause contamination and damage the internal mechanism.
Shop By Brand