Methanol
Methanol
Methanol is a widely used laboratory solvent valued for its high polarity, volatility and compatibility with analytical techniques. In research, quality control and routine laboratory workflows, methanol plays a critical role in chromatography, sample preparation and chemical synthesis.
At the Fisher Scientific channel, we offer a comprehensive range of methanol grades, including HPLC, LC-MS and analytical reagent qualities. Sourced from trusted manufacturers such as Fisher Chemicals, Honeywell Fluka, Honeywell Research Chemicals and JT Baker, these products deliver consistent purity, low UV absorbance and reliable performance across applications.
Methanol, Optima™ LC/MS Grade, Fisher Chemical
Methanol, >-99.9%, CH4O, CAS Number-67-56-1, wood alcohol, methyl hydroxide, colonial spirit, columbian spirit, wood naphtha, carbinol, methyl alcohol, pyroxylic spirit, wood spirit, methylol | CAS: 67-56-1 | CH4O | 32.04 g/mol
    Methanol, Optima™ LC/MS Grade, Fisher Chemical
    Methanol, Certified AR for Analysis, Fisher Chemical™
    CH4O, CAS Number-67-56-1, wood naphtha, colonial spirit, pyroxylic spirit, methyl hydroxide, methyl alcohol, wood spirit, wood alcohol, methylol, columbian spirit, carbinol, 2.5L, 64.7 deg.C, CHEBI:17790, Colorless, 32.04g/mol, OKKJLVBELUTLKV-UHFFFAOYSA-N, methanol, -98 deg.C, 4595, 32.042 | CAS: 67-56-1 | CH4O | 32.04 g/mol
      Corning™ Costar™ Spin-X™ Centrifuge Tube Filters, CA Membrane
      Methanol CHROMASOLV™, for HPLC, ≥99.9%, Honeywell Riedel-de Haën™
      Product filtered through a 0.2μm filter
      • For HPLC and organic synthesis
      • High-purity, multipurpose solvents
      • High lot-to-lot consistency
        Methanol CHROMASOLV™, for HPLC, ≥99.9%, Honeywell Riedel-de Haën™
        Save Now - Exclusive Deals
        Complete Your Order - Great Deals
        Brady™ M210-LAB Label Printer, Carton Pack
        The M210-LAB Portable Label Printer is an all-in-one labeler for research, academic and clinical labs, creating legible labels for flat, curved or highly textured surfaces that stay stuck for years- even when exposed to extreme temperatures and chemicals.
        Brady™ M210-LAB Label Printer- Carton Pack
        Fisherbrand™ Squat Form Beakers
        Constructed of borosilicate glass. Fisherbrand™ Squat Form Beakers are graduated and come in a large variety of capacities.
        Fisherbrand™ Squat Form Beakers
        Fisherbrand™ Visitorspec Safety Glasses, Coated, Antifog
        Ideal for protecting visitors in industrial or medical settings.
        • Economical safety spectacles
        • 100% impact-resistant polycarbonate wraparound unilens design with blue temples
        • Clear lens absorbs >99.9% of harmful UV light
        • Fit over prescription eyeglasses
        • Scratch-resistant, Anti-fog, Anti-static
        Fisherbrand™ Visitorspec Safety Glasses, Coated, Antifog
        Must Have
        FAQ
        Methanol is used as a solvent for HPLC, LC-MS, sample preparation and synthesis. High-purity grades from Fisher Chemical and Honeywell ensure reproducible analytical results.
        Choose LC-MS or HPLC methanol for chromatography and analytical reagent grade for routine analysis. JT Baker and Honeywell offer certified options for specific methods.
        Fisher Chemical, Honeywell Fluka, Honeywell Research Chemicals and JT Baker are leading suppliers of high-purity methanol for laboratory use.
        LC-MS methanol has ultra-low impurities that reduce background noise and improve sensitivity, supporting accurate mass spectrometry analysis.
        Methanol should be stored in approved flammable storage cabinets, tightly sealed and clearly labelled, following laboratory safety guidelines.
        Top Brands