Microplate Readers and Washers
Microplate Readers and Washers
Discover Thermo Scientific™ Detection Solutions.
If you are searching for microplate readers or washers, explore the Thermo Scientific™ portfolio — a comprehensive range of reliable, high-performance systems for ELISA, cell-based assays, nucleic acid quantification, protein analysis, and advanced research applications.
Available through the Fisher Scientific channel with full pricing visibility, service support and application expertise.
Multimode Microplate Readers
Flexible detection for evolving research needs.
Thermo Scientific multimode microplate readers provide powerful, configurable detection technologies to support a wide range of assays — from routine fluorescence measurements to advanced luminescence and alpha-based applications.
Choose the right system for your workflow:
Thermo Scientific™ Varioskan™ LUX Multimode Microplate Reader
Advanced, fully configurable multimode detection for research applications.
A highly flexible, modular platform supporting absorbance, fluorescence, luminescence, time-resolved fluorescence (TRF), and AlphaScreen™ technologies. Designed for laboratories requiring maximum sensitivity, flexibility, and future scalability.
Best suited for:
  • Advanced assay development
  • TRF and AlphaScreen™ applications
  • Cell-based assays and kinetic workflows
  • Labs requiring dispensers, gas control or bottom reading
Key benefits across the range:
  • Broad assay compatibility
  • Upgradeable configurations (model dependent)
  • Intuitive SkanIt™ software
  • Suitable for academic, biotech, and pharmaceutical research
Thermo Scientific™ Varioskan™ LUX Multimode Microplate Reader
Thermo Scientific™ Varioskan™ ALF Multimode Microplate Reader
Reliable, cost-effective multimode detection for routine applications.
A filter-based multimode reader combining absorbance, fluorescence, and luminescence in a compact, easy-to-use system. Ideal for laboratories running established assays and standard workflows.
Best suited for:
  • Routine fluorescence and luminescence assays
  • ELISA and basic research workflows
  • Budget-conscious labs needing multimode capability
Key benefits across the range:
  • Broad assay compatibility
  • Upgradeable configurations (model dependent)
  • Intuitive SkanIt™ software
  • Suitable for academic, biotech, and pharmaceutical research
Thermo Scientific™ Varioskan™ ALF Multimode Microplate Reader
Absorbance Microplate Readers
Robust performance for ELISA and routine quantification.
Thermo Scientific absorbance microplate readers deliver dependable, reproducible performance for ELISA workflows and general absorbance-based assays.
Choose the right system for your workflow:
Thermo Scientific™ Multiskan™ SkyHigh Microplate Spectrophotometer
Advanced absorbance detection with enhanced flexibility.
A monochromator-based microplate spectrophotometer offering expanded wavelength selection, optional, touchscreen operation and optional cuvette capability. Designed for laboratories requiring greater flexibility and broader assay compatibility.
Best suited for:
  • Multi-assay environments requiring wavelength flexibility
  • DNA/RNA and protein quantification in the UV range
  • Laboratories needing cuvette measurements
  • Users seeking a modern interface and expanded functionality
Key benefits across the range:
  • Reliable, reproducible absorbance measurements
  • Optimised for ELISA and biochemical assays
  • Intuitive SkanIt™ software for streamlined workflows
  • Scalable solutions to match application needs and budget
Thermo Scientific™ Multiskan™ SkyHigh Microplate Spectrophotometer
Thermo Scientific™ Multiskan™ FC Microplate Photometer
Simple, reliable absorbance detection for routine ELISA workflows.
A compact, filter-based microplate photometer designed for straightforward, high-confidence absorbance measurements. Ideal for laboratories running established ELISA assays and for routine testing.
Best suited for:
  • Routine ELISA workflows
  • Endpoint absorbance measurements
  • Quality control and teaching laboratories
  • Users prioritising simplicity and cost-efficiency
Key benefits across the range:
  • Reliable, reproducible absorbance measurements
  • Optimised for ELISA and biochemical assays
  • Intuitive SkanIt™ software for streamlined workflows
  • Scalable solutions to match application needs and budget
Thermo Scientific™ Multiskan™ FC Microplate Photometer
Automated Microplate Washers
Consistent, reliable washing for ELISA and cell-based assays.
Thermo Scientific microplate washers provide precise, programmable washing performance to support reproducibility and assay accuracy.
Thermo Scientific™ Wellwash™ Microplate Washer
Reliable, easy-to-use washing for routine ELISA applications.
A robust washer designed for consistent performance in standard ELISA workflows. Offers simple operation and dependable results for laboratories focused on routine testing.
Best suited for:
  • Routine ELISA washing
  • Standard 96-well plate workflows
  • Laboratories prioritising ease of use and reliability
  • Cost-conscious environments
Key benefits across the range:
  • Consistent and reproducible washing performance
  • Adjustable wash parameters for different assay needs
  • Reduced residual volumes to improve assay accuracy
  • User-friendly operation for efficient workflows
Thermo Scientific™ Wellwash™ Microplate Washer
Thermo Scientific™ Wellwash™ Versa Microplate Washer
Flexible washing for higher throughput and more demanding workflows.
An advanced washer supporting both 96-well and 384-well plates with enhanced programmability and flexibility. Designed for laboratories handling more complex assays or higher throughput requirements.
Best suited for:
  • 96- and 384-Well plate formats
  • Higher throughput workflows
  • More complex or customised wash protocols
  • Laboratories requiring greater flexibility and control
Key benefits across the range:
  • Consistent and reproducible washing performance
  • Adjustable wash parameters for different assay needs
  • Reduced residual volumes to improve assay accuracy
  • User-friendly operation for efficient workflows
Thermo Scientific™ Wellwash™ Versa Microplate Washer
FAQ
Step 1: What detection mode do you require?
  • Absorbance only → Explore Multiskan™ FC or Multiskan™ SkyHigh
  • Absorbance + fluorescence + luminescence → Explore Varioskan™ ALF
  • Advanced detection including TRF or AlphaScreen™ → Explore Varioskan™ LUX
Step 2: Do you require reagent dispensers or gas control?
  • Yes → Varioskan™ LUX
  • No → Varioskan™ ALF or Multiskan™ models
Step 3: Are you running cell-based assays with adherent cells?
  • Yes → Consider Varioskan™ LUX (bottom reading capability)
Thermo Scientific microplate readers and washers are trusted worldwide for their reliability, performance and ease of integration into laboratory workflows.
  • Comprehensive Portfolio
    From routine absorbance measurements to advanced multimode detection
  • Scalable Solutions
    Systems suitable for teaching labs, research institutes, and high-throughput environments
  • Intuitive Software
    SkanIt™ software enables straightforward assay setup, data acquisition, and analysis
  • Service and Support
    Backed by global service infrastructure with service contracts available through the Fisher Scientific channel