Microplate Sealing Films and Tapes
Achieve reliable PCR, qPCR, cell culture, and long-term storage results with microplate sealing films and tapes from leading brands. Compatible with 96- and 384-well plates, these low-autofluorescence and solvent-resistant options help prevent evaporation, contamination, and edge effects while supporting automation and high-throughput screening.
Applied Biosystems™ MicroAmp™ Optical Adhesive Film
Reduces chance of well-to-well contamination and sample evaporation when applied to a microplate.
Applied Biosystems™ MicroAmp™ Optical Adhesive Film
Thermo Scientific™ Adhesive PCR Plate Seals
Provides an effective and convenient sealing method for PCR applications.
Thermo Scientific™ Adhesive PCR Plate Seals
Thermo Scientific™ Adhesive PCR Plate Foils
Strong adhesive prevents evaporation during thermal cycling or sample storage.
Thermo Scientific™ Adhesive PCR Plate Foils
Save Now - Exclusive Deals
Complete Your Order - Great Deals
Applied Biosystems™ MicroAmp™ Adhesive Film Applicator
Use to form a secure seal between a microplate and an adhesive film.
Applied Biosystems™ MicroAmp™ Adhesive Film Applicator
Nucleic Acid Reaction Assay Microplates
Plates for PCR, qPCR, DNA or RNA reactions designed to fit numerous thermal cycler models and instruments that sustain enzymatic and biochemical reactions. Choose from plates with or without barcodes, flat, U- or V-shaped bottoms, and varying sterility and volume parameters depending on your needs.
Nucleic Acid Reaction Assay Microplates
Storage Microplates
Multi-well plates (typically polypropylene with deep wells), designed for short- or long-term sample storage or incubation; includes numerous formats (well numbers, shapes, sealable) and qualities (sterile, non-pyrogenic, DNase-free, RNase-free).
Storage Microplates
FAQ
Microplate sealing options include optically clear adhesive films for qPCR, aluminium foils for thermal cycling, gas-permeable films for cell culture and robust storage tapes. Brands such as Applied Biosystems, Thermo Scientific Nunc, Thermo Scientific Matrix, Corning, Axygen and Brand provide options for 96-well and 384-well plates.
Optical adhesive films are recommended for qPCR and fluorescence-based assays where low background and clear signal are critical. Aluminium foils provide strong evaporation control and chemical resistance for PCR, storage and transport. Selection depends on detection method, temperature range and whether plates require piercing.
For long-term storage in freezers or cold rooms, robust aluminium foils and specialised storage tapes from Thermo Scientific, Corning, Axygen and Brand offer strong adhesion and barrier properties. These products help reduce sample evaporation, preserve concentration and protect sensitive biomolecules during repeated temperature cycling.
Effective microplate sealing films and tapes minimise edge effects, evaporation and cross-contamination, which are common issues in high-throughput screening. Using high-quality seals from Applied Biosystems, Thermo Scientific Nunc, Thermo Scientific Matrix, Corning and other brands supports consistent assay conditions and reproducible data across entire plate runs.
The microplate sealing films and tapes category includes seals compatible with a wide variety of 96-well and 384-well plate designs. By filtering by brand, material or application, you can match Applied Biosystems, Thermo Scientific, Corning, Axygen and Brand sealing products to your current plate formats and automation setup.
Top Brands