PCR Tubes
PCR Tubes
PCR tubes are essential consumables for accurate and efficient thermal cycling workflows in molecular biology laboratories. The Fisher Scientific channel offers a broad portfolio of PCR tubes designed for consistent heat transfer, reliable sealing, and compatibility with leading thermal cyclers. Browse trusted brands including Thermo Scientific, Applied Biosystems, Invitrogen, Fisherbrand, Eppendorf, Axygen, Brand, and MP Biomedicals to support PCR, qPCR, and routine sample preparation applications.
Eppendorf™ PCR Clean Colorless Safe-Lock Centrifuge Tubes
Protect your samples with these centrifuge tubes. Eppendorf™ PCR Clean Colorless Safe-Lock Centrifuge Tubes feature a hinged Eppendorf Safe–Lock lid for outstanding protection against unintentional opening during incubation and storage.
Eppendorf™ PCR Clean Colorless Safe-Lock Centrifuge Tubes
Eppendorf™ PCR Clean Colorless Safe-Lock Centrifuge Tubes
Protect your samples with these centrifuge tubes. Eppendorf™ PCR Clean Colorless Safe-Lock Centrifuge Tubes feature a hinged Eppendorf Safe–Lock lid for outstanding protection against unintentional opening during incubation and storage.
Eppendorf™ PCR Clean Colorless Safe-Lock Centrifuge Tubes
Fisherbrand™ 0.2mL PCR Tubes
Cleanroom produced from virgin polypropylene, featuring a thin-wall design with integral snap-shut cap.
Fisherbrand™ 0.2mL PCR Tubes
Save Now - Exclusive Deals
Complete Your Order - Great Deals
Electronic Multichannel Pipettes
Our range of electronic multichannel pipettes are used with disposable pipette tips to simultaneously transfer multiple measured amounts of liquid between various labware formats.
Electronic Multichannel Pipettes
Manual Multichannel Pipettes
Our range of manual multichannel pipettes are designed to simultaneously transfer multiple batches of liquid between labware of different formats. Contamination is avoided using disposable pipette tips.
Manual Multichannel Pipettes
Universal Pipette Tips
Universal pipette tips are compatible with pipettes from various manufacturers and have comparable performance to branded tips. Suitable for any genomic or biological application, they are available in sterile and non-sterile versions.
Universal Pipette Tips
FAQ
PCR tubes are used to hold reaction mixtures during polymerase chain reaction workflows. They are designed to withstand repeated heating and cooling cycles while maintaining sample integrity and consistent thermal transfer for reliable amplification results.
Thin-wall PCR tubes improve heat transfer efficiency during thermal cycling. This supports faster temperature equilibration and more consistent amplification, helping laboratories achieve reliable PCR and qPCR performance across routine workflows.
Leading PCR tube brands available from the Fisher Scientific channel include Eppendorf, Thermo Scientific, Fisherbrand, Axygen, Brand, Invitrogen, Applied Biosystems, and MP Biomedicals for molecular biology and research applications.
PCR tube compatibility is important because different thermal cyclers require specific dimensions and sealing formats. Compatible PCR consumables help ensure accurate thermal contact, minimise evaporation, and support reproducible experimental outcomes.
Laboratories can purchase PCR tubes in bulk from the Fisher Scientific channel through a wide range of packaging formats and trusted laboratory brands designed to support high-throughput molecular biology workflows and routine PCR testing.
Top Brands