Protein Mass Spectrometry Sample Preparation
Protein Mass Spectrometry Sample Preparation
Prepare confident, publication-ready proteomics samples with curated Protein Mass Spectrometry Sample Preparation essentials. From calibration solutions and digestion standards to MS-grade proteases, peptide cleanup, and enrichment tools, this selection helps improve identification, quantitation, and run-to-run consistency. Shop trusted options from Thermo Scientific, Invitrogen, and Cytiva to support robust LC-MS workflows across discovery and targeted proteomics.
Thermo Scientific™ Pierce™ FlexMix™ Calibration Solution
Thermo Scientific™ Pierce™ FlexMix™ Calibration Solution is a mixture of 16 highly pure, ionizable components designed for both positive and negative ionization calibration using Orbitrap Tribrid Series 3.1 Instrument Control Software.
      Thermo Scientific™ Pierce™ FlexMix™ Calibration Solution
      Thermo Scientific™ Pierce LTQ Velos ESI Positive Ion Calibration Solution
      Thermo Scientific™ Pierce LTQ Velos ESI Positive Ion Calibration Solution is a mixture of highly purified ionizable molecules designed for positive mode calibration of Thermo Scientific™ LTQ Velos series mass spectrometer instruments.
        Thermo Scientific™ Pierce™ LTQ Velos ESI Positive Ion Calibration Solution
        Cytiva HiTrap™ IMAC Columns
        For optimizing purification of histidine-tagged proteins when Ni+2 is not the best choice of metal ion
          Cytiva HiTrap™ IMAC Columns
          Complete Your Order - Great Deals
          Thermo Scientific™ aminoxyTMTsixplex™ Label Reagent Set
          The carbonyl-reactive Thermo Scientific™ aminoxyTMT™ (Tandem Mass Tag™) Label Reagents enable multiplexed characterization and quantitation of carbonyl-containing biomolecules (carbohydrates, steroids, oxidized proteins) by mass spectrometry (MS).
          Thermo Scientific™ aminoxyTMTsixplex™ Label Reagent Set
          Thermo Scientific™ Pierce Dimethylsulfoxide (DMSO)
          High-purity, sequencing-grade DMSO engineered for reliable HPLC and mass spectrometry performance — delivering consistent absorptivity and confidence in every run.
          Thermo Scientific™ Pierce™ Dimethylsulfoxide (DMSO), LC-MS Grade
          Acetonitrile, Optima™ LC/MS Grade, Fisher Chemical™
          Acetonitrile, >-99.9%, C2H3N, CAS Number-75-05-8, methane, cyano, ethyl nitrile, methanecarbonitrile, methylkyanid, mecn, acetonitrile, methyl cyanide, ethanenitrile, cyanure de methyl, cyanomethane, 1L | CAS: 75-05-8 | C2H3N | 41.053 g/mol
          Acetonitrile, Optima™ LC/MS Grade, Fisher Chemical™
          FAQ
          Protein MS sample preparation is the set of steps that converts complex protein mixtures into clean, MS-compatible peptides. Typical workflows include protein quantitation, reduction/alkylation, enzymatic digestion (e.g., trypsin or Lys-C), cleanup/desalting, and optional enrichment. Products from Thermo Scientific, Invitrogen, and Cytiva support consistent LC-MS performance.
          Trypsin is a standard protease for broad peptide coverage and predictable cleavage, while Lys-C is often used to improve digestion robustness, especially under denaturing conditions. Many labs use Lys-C first, followed by trypsin, to increase completeness and reproducibility. Thermo Scientific offers MS-grade options commonly used in LC-MS workflows.
          The biggest gains typically come from removing salts, detergents, and other ion-suppressing contaminants, plus controlling digestion and calibration. Peptide cleanup (C18 tips/spin columns), consistent standards, and reliable quantitation help stabilize run-to-run results. Thermo Scientific and Invitrogen solutions support tuning and QC; Cytiva tools can support enrichment and cleanup needs.
          Missed cleavages often drop when you optimize denaturation, maintain correct enzyme-to-protein ratios, control incubation time/temperature, and remove inhibitors (e.g., detergents). Using MS-grade proteases and compatible surfactants can improve digestion efficiency. Pairing careful quantitation with consistent Thermo Scientific digestion reagents helps standardize outcomes across experiments.
          Peptide cleanup (often C18-based) removes salts, buffers, and small molecules that suppress ionization and contaminate LC-MS systems. Clean peptides typically yield better sensitivity, fewer background ions, and more stable chromatography. Thermo Scientific cleanup consumables are widely used for desalting and concentration steps before analysis.
          Calibration solutions help verify and adjust instrument mass accuracy, while standards and digests provide routine checks on sensitivity, chromatography, and identification performance. Together, they enable trend monitoring across sequences and instruments. Invitrogen and Thermo Scientific calibration solutions, along with reference digests, support comparability and confident troubleshooting.
          Phosphopeptide enrichment concentrates modified peptides to improve detection and site localization. Common approaches include TiO2-based enrichment and IMAC workflows, followed by careful cleanup to reduce nonspecific binding. Thermo Scientific enrichment kits and Cytiva chromatography options can support targeted enrichment strategies, especially when combined with optimized buffers and loading conditions.
          Top Brands