Real-Time PCR Assays and Arrays
Real-Time PCR Assays and Arrays are powerful tools for detecting and quantifying DNA or RNA sequences with high precision. These techniques enable real-time monitoring of nucleic acid amplification and allow for the simultaneous analysis of multiple genes, making them essential in diagnostics, gene expression studies, and various research applications.
TaqMan™ Gene Expression Assay (FAM)
Used for quantitative real-time PCR analysis of gene expression.  Each assay includes target primers and a sequence-specific probe optimized for the best functional performance.
TaqMan™ Gene Expression Assay (FAM)
TaqMan™ Gene Expression Assay, VIC
The gold standard in real-time PCR gene expression studies.
TaqMan™ Gene Expression Assay, VIC
Thermo Scientific™ RapidFinder™ Ruminant ID Kit
Simple and reliable detection in food and feed samples.
Thermo Scientific™ RapidFinder™ Ruminant ID Kit
Complete Your Order - Great Deals
Nucleic Acid Reaction Assay Microplates
Plates for PCR, qPCR, and DNA/RNA reactions compatible with many thermal cyclers. Available with or without barcodes, in flat, U-, or V-bottom formats, and in various sterility and volume options to suit your needs.
Nucleic Acid Reaction Assay Microplates
PCR Tubes
PCR tubes are small tubes for PCR applications, offered in various materials, volumes, and RCF ratings. They may be autoclavable and RNase-, DNase-, and DNA-free, and come in multiple colors for easy sample identification. Available as individual tubes or strips.
PCR Tubes
Thermal Cyclers
Explore our advanced thermal cyclers for temperature-sensitive applications like PCR. Choose the temperature range, capacity, and accessories you need to find the ideal fit for your lab.
Thermal Cyclers
FAQ
Real-time PCR (qPCR) is a technique used to amplify and quantify DNA in real-time. Key points include:
  • Sample Preparation: Extract DNA from the sample.
  • Reaction Setup: Prepare a mixture with DNA template, primers, DNA polymerase, nucleotides, and fluorescent dyes/probes.
  • Thermal Cycling: Repeated cycles of heating and cooling to denature DNA, anneal primers, and extend new DNA strands.
  • Fluorescence Detection: Measure fluorescence emitted by dyes/probes binding to DNA, proportional to the amount of DNA amplified.
  • Data Analysis: Generate an amplification curve to determine the initial quantity of target DNA using the cycle threshold (Ct) value.
Real-time PCR is widely used in various applications, including:
  • Medical diagnostics: Detecting and quantifying pathogens (e.g., viruses, bacteria).
  • Genetic testing: Identifying genetic mutations or variations.
  • Research: Studying gene expression levels.
  • Forensics: Analyzing DNA samples for identification purposes.
The key advantage of real-time PCR is its ability to provide quantitative data with high sensitivity and specificity.
Common challenges in utilizing real-time PCR assays and arrays, along with their solutions, include:
  • Non-Specific Amplification: non-specific products or primer-dimer formations can interfere with accurate quantification. To overcome this, design primers carefully to ensure specificity, optimize annealing temperatures, and use hot-start DNA polymerases.
  • Inconsistent Results: variability in results due to inconsistent sample preparation or pipetting errors. To avoid this, use automated pipetting systems, ensure thorough mixing of reagents, and include internal controls to monitor consistency.
  • Low Sensitivity: difficulty detecting low-abundance targets. To overcome this, optimize reaction conditions, use high-quality reagents, and ensure efficient extraction and purification of nucleic acids.
  • Inhibition of PCR Reaction: presence of inhibitors in the sample that affect the PCR efficiency. A solution to this is to use purification steps to remove inhibitors, include an internal control to detect inhibition, and use inhibitor-resistant enzymes.
  • Data Analysis and Interpretation: complex data analysis and potential for misinterpretation of results. Utilize robust software for data analysis, ensure proper training for users, and validate results with replicates and appropriate controls.
When buying real-time PCR assays and arrays, consider the following five key factors:
  1. Sensitivity and Specificity: Ensure the assay can detect low levels of target DNA/RNA with high specificity to avoid false positives and negatives.
  2. Compatibility: Verify that the assays and arrays are compatible with your real-time PCR instrument and software.
  3. Validation and Quality Control: Look for assays that are validated and come with quality control data to ensure reliable and reproducible results.
  4. Cost and Budget: Consider the cost per assay and overall budget, including any additional reagents or consumables required.
  5. Technical Support and Documentation: Ensure the supplier provides comprehensive technical support, user manuals, and troubleshooting guides.
Resources and Related Information
Should You Use Digital or Real-Time PCR?
Real-time PCR vs. traditional PCR vs. digital PCR at a glance.
Should You Use Digital or Real-Time PCR?
TaqMan Assays for Genetic Variation Research
Learn how TaqMan Assays can help in analyzing genetic variation in animals and plants.
TaqMan Assays for Genetic Variation Research
Taqman Gene Expression Assay Solutions
Find information on the TaqMan Gene Expression Assays - the gold standard in performance and quality, for gene expression analysis.
Taqman Gene Expression Assay Solutions
Top Brands