Round Plastic Bottles
Round plastic bottles are a core choice for liquid storage, sample collection and transport across chemistry, life science and environmental labs. This range from Thermo Scientific Nalgene, Fisherbrand, Azlon, Kautex and Gosselin covers volumes from small sample vials to multi-litre containers in HDPE and LDPE materials. Options include wide-neck designs for easy filling, leak-resistant closures, graduated walls and robust construction for demanding workflows. Use these bottles for buffer and media storage, reagent preparation, waste collection and shipping of water or soil samples while supporting consistent, reproducible results across sites and teams.
Kautex™ LDPE Wide Mouth Bottles Series 303
Designed with a wide opening for easy filling and cleaning. Kautex™ Wide Mouth Bottles are ideal for liquids, pastes and granules.
Contec™ Sterile IPA
Thermo Scientific™ Nalgene™ Wide-Mouth Lab Quality HDPE Bottles
Perform countless field applications utilizing these general-purpose, wide-mouth, lab quality HDPE bottles.
Contec™ Sterile IPA
Save Now - Exclusive Deals
FAQ
Most round plastic bottles in this range are manufactured from HDPE or LDPE. HDPE bottles from Thermo Scientific Nalgene, Fisherbrand and Azlon provide good chemical resistance and rigidity, while LDPE bottles from Kautex and Gosselin offer improved flexibility and reliable squeeze control.
Select bottle volume according to the working volume you need plus headspace for expansion, mixing or freezing. Smaller Fisherbrand and Gosselin bottles help minimise reagent waste, while larger Thermo Scientific Nalgene and Azlon containers suit bulk buffer storage or transport of environmental samples.
Round plastic bottles are available with standard screw caps, wide-mouth leak-resistant caps and specialised pouring or dispensing closures. Thermo Scientific Nalgene, Fisherbrand, Azlon, Kautex and Gosselin offer compatible caps designed to maintain a secure, reliable seal during storage and transport.
Check the chemical compatibility of HDPE or LDPE with your solvent or reagent using manufacturer data from Thermo Scientific Nalgene, Azlon or Kautex. For aggressive chemicals, verify maximum concentration, temperature limits and contact times to avoid swelling, cracking or permeation of the bottle walls.
Branded round plastic bottles from Thermo Scientific Nalgene, Fisherbrand, Azlon, Kautex and Gosselin typically offer better batch consistency, traceability and validated performance. This supports standardised storage procedures, reproducible results and a lower risk of leaks or contamination during transport and handling.
Top Brands