Serological Pipettes
Serological pipettes are used for transferring liquids and mixing chemical solutions or cell suspensions with the help of a pipette dispenser. Offered in graduated glass or plastic and marked 'To Contain' or 'To Deliver', they are available in both sterile or non-sterile forms and different lengths.
EASY-TO-READ
Sterile polystyrene disposable serological pipets
Precision and clarity in every measurement.
Contec™ NeutraKlean™ Detergent with Purified Water, Sterile
READY-TO-USE
Stripette™ Paper-Wrapped Disposable Polystyrene Serological Pipettes
Reliable and convenient for everyday use.
Contec™ Non-Sterile CyChlor
Complete Your Order - Great Deals
Fisherbrand™ Cargo Pipette Racks
Store and organize your 1mL to 50mL serological pipettes.
Universal Pipette Tips
Thermo Scientific™ S1 Pipette Fillers
Ergonomically designed pipette fillers for use with graduated and volumetric glass or plastic serological pipettes.
Pipette Specific Tips
Cell Culture Dishes
Browse through a wide range of cell culture dishes.
Pipette Specific Tips
FAQ
Serological pipets offer several benefits in a laboratory setting, including versatility and precision in liquid handling. They are ideal for measuring and transferring larger volumes of liquids, making them suitable for various applications such as cell culture, reagent preparation, and sample mixing. Serological pipets are typically graduated, allowing for accurate measurement of liquid volumes. They are available in different sizes and materials, such as plastic or glass, to accommodate specific experimental needs. Additionally, serological pipets are designed for single-use or reusable purposes, providing flexibility and convenience in the lab.
Proper use and disposal of serological pipets are essential for maintaining laboratory safety and accuracy. When using serological pipets, ensure that you select the appropriate size and material for your application. Attach the pipet to a pipet filler or controller for precise liquid handling. Avoid touching the pipet tip to prevent contamination. After use, dispose of single-use pipets in designated biohazard or waste containers according to your laboratory's protocols. For reusable pipets, clean and sterilize them according to the manufacturer's instructions before reuse. Proper handling and disposal practices will help maintain a safe and efficient laboratory environment.
Common issues with serological pipets include inaccurate volume measurement, which can be caused by improper handling or using a damaged pipet. Ensure that you are using the correct pipet size and handling it properly to achieve accurate measurements. Contamination is another issue, especially when pipets are reused; always follow proper sterilization procedures to prevent contamination. Leaking can occur if the pipet is damaged or if it is not securely attached to the pipet filler. Regularly inspect pipets for any signs of damage and ensure proper attachment to the pipet filler. By addressing these common issues through proper selection, handling, and maintenance, you can ensure the reliable performance of your serological pipets.
Shop By Brand