Surface Binding Assay Microplates
Surface Binding Assay Microplates
Assay plates with different surface treatments and binding types promote non-specific surface adsorption of antibodies, proteins or other biomolecules for quantitative assays such as ELISA. Choose sterile, non-sterile, flat, U- or V-shaped bottoms, different arrays and colors depending on your need.
FEATURED
Thermo Sceintific™ Clear Flat Bottom Immuno Nonsterile 96-Well Plates
Choose from four binding surfaces of these 96-well plates for reliable and reproducible results in colometric assays.
Contec™ Sterile IPA
BINDING A DIVERSE RANGE
Thermo Scientific™ Black 96-Well Immuno Plates
Choose from PolySorp, MaxiSorp, and the Microfluor 2/2+ plate surfaces for binding a diverse range of biomolecules.
Contec™ Sterile IPA
FOR MOST APPLICATIONS
BRAND™ BRANDplates™ pureGrade™ 384-Well Microplates
Non-treated PS surface, medium binding, non-sterile.
Contec™ Sterile IPA
Complete Your Order - Great Deals
FAQ
Surface binding assay microplates are used for immobilizing biomolecules such as proteins, nucleic acids, or cells to the surface of the wells, enabling various types of assays including ELISA, cell-based assays, protein-protein interaction studies, and nucleic acid hybridization assays.
Different well shapes in surface binding assay microplates are optimized for various methods and applications. Choosing the appropriate well shape ensures optimal performance.
Flat-Bottom Wells: Ideal for absorbance-based assays (e.g., ELISA) and cell culture. Reason: provides a uniform surface for cell attachment and consistent optical path length for absorbance measurements.
Round-Bottom Wells: Suitable for suspension cell culture, mixing, and centrifugation. Reason: facilitates better mixing and pelleting of cells or beads during centrifugation.
V-Bottom Wells: Best for assays requiring efficient sample recovery, such as pelleting cells or beads.
U-Bottom Wells: Commonly used for agglutination assays and certain types of cell culture. Reason: The rounded shape aids in visualizing agglutination and allows cells to settle in a consistent pattern.
When selecting Surface Binding Assay Microplates, consider the following five factors:
1) The surface chemistry of the microplate should be compatible with the biomolecules you intend to bind. Different coatings (e.g., polystyrene, glass, or specialized coatings like streptavidin) offer varying binding affinities for proteins, nucleic acids, or cells.
2) Choose the appropriate well format (e.g., 96-well, 384-well, or 1536-well plates) based on the throughput requirements of your assay.
3) Ensure the microplate has suitable optical clarity and minimal background fluorescence or absorbance for your detection method (e.g., fluorescence, absorbance, or luminescence).
4) Select a microplate with a binding capacity that matches your assay's needs to ensure optimal performance.
5) Verify that the microplate is compatible with the instruments used for detection and analysis in your lab.
Resources and Related Information
Corning Life Sciences Selection Guide
For superior quality and performance in life science research tools, you can count on Corning. Our comprehensive line of advanced products and technologies is designed to meet your evolving needs in cell-based and genomics research.
Protective Solutions For Pharmaceutical Industry Applications
Multiserotype Enzyme-Linked Immunosorbent Assay using Microtiter Cliniplate
The aim of the study was to measure serum immunoglobulin G class antibodies against Actinobacillus actinomycetemcomitans and Porphyromonas gingivalis by an enzyme-linked immunosorbent assay (ELISA) in which mixtures of several serotypes of the pathogens were used as antigens to avoid biasing of the results in favor of a particular strain.
Multiserotype Enzyme-Linked Immunosorbent Assay using Microtiter Cliniplate
ELISA Guide
A clear and easy guide to ELISAs
ELISA Guide
Shop By Brand