Thermal Cyclers
Thermal Cyclers
Choosing the right thermal cycler (also known as a thermocycler or PCR machine) is key to the success of your PCR experiments across many applications, including sequencing, genotyping, cloning, and mutagenesis. Applied Biosystems™  thermal cyclers help generate precise, consistent, reproducible results for every challenge, experiment, and budget.
Applied Biosystems SimpliAmp Thermal Cycler
The SimpliAmp™ Thermal Cycler is a small, easy-to-use, and accurate thermal cycler that suits any lab's needs for everyday PCR.
Contec™ Sterile IPA
Applied Biosystems VeritiPro Thermal Cycler, 96-Well
The VeritiPro™ Thermal Cycler delivers proven reliability with advanced temperature control technology and connectivity.
Contec™ Sterile IPA
Applied Biosystems PowerFlex PCR System, 96-Well & 32-Well
Meet the next-generation PowerFlex™ Thermal Cycler. Built for powerfully precise and flexible PCR workflows of any application.
  • Performance: Accuracy in temperature gradient precision
  • Compatibility: Seamless integration with PCR plastics formats
  • Productivity: Maximize the efficiency of your lab with minimal downtime
Applied Biosystems™ PowerFlex™ Thermal Cyclers
Save Now - Exclusive Deals
Must Have
Complete Your Order - Great Deals
PCR Tubes
PCR tubes are small tubes designed for use in PCR applications. Available in a variety of materials, volumes, and RCF ratings, they may be autoclavable and/or RNase-, DNase-, and DNA-free.
Lorem Ipsum
Real Time PCR Master Mixes
Real-time PCR, or qPCR, permits the detection and quantification of target nucleic acids. PCR master mixes can be used to reduce the risk of contamination and other errors related to mixing and pipetting.
Lorem Ipsum
Hot Start PCR Reagents and Kits
Reduce non-specific DNA amplification that occurs naturally at lower temperatures with our broad range of hot start PCR products. Browse through kits, master mixes, polymerases, and buffers to obtain increased yield, specificity, and sensitivity for all of your PCR application needs.
Lorem Ipsum
FAQ
A thermal cycler, often called a PCR machine, is a laboratory instrument used to amplify DNA through the polymerase chain reaction (PCR) process. It automates the repeated heating and cooling cycles needed for DNA denaturation, primer annealing, and strand extension, making it essential in molecular biology, genetics, diagnostics, and forensic science.
A thermal cycler improves PCR efficiency and accuracy by maintaining precise and consistent temperature control throughout each cycle. This ensures reliable DNA amplification with minimal errors. Advanced thermal cyclers include features such as gradient temperature settings, fast heating and cooling ramp rates, and programmable protocols, which improve PCR specificity, reproducibility, and yield for applications like genetic testing, cloning, and disease detection.
When selecting the best thermal cycler for PCR, consider these key features:
  • Precise temperature control for consistent results
  • Fast ramp rates to reduce cycle time
  • User-friendly interface and programing options
  • Capacity for multiple samples (PCR tubes, strips, or 96-well plates)
  • Gradient functionality for optimizing annealing temperatures
  • Compatibility with standard PCR consumables
These features help researchers choose a thermal cycler that matches their PCR application needs, whether in academic research, clinical diagnostics, or industrial biotechnology.
Top Brands