Ultra Low Temperature Freezers
Ultra Low Temperature Freezers
Ultra-low temperature freezers ensure safe, long-term storage of biological and chemical samples at up to −80°C. Used in research labs, biobanks, pharma, and clinical settings, our range from Thermo Scientific, Eppendorf, Fisherbrand, and Binder supports strict storage, compliance, and energy-efficiency needs across Europe.
Thermo Scientific™ TDE Series -86°C Ultra-Low Temperature Freezers
The TDE Series is a sustainable and reliable solution with ultimate sample security and operational savings for every lab. Meets ±10°C validation protocols at -80°C with strong peak variation and steady-state temperature variation.
Thermo Scientific™ TDE Series -86°C Ultra-Low Temperature Freezers
Thermo Scientific™ TSX Universal Series General Purpose Ultra-Low Freezers
The next generation of our most trusted ULT platform adapts to your needs, reduces your environmental impact, and is built on our most robust design ever
Thermo Scientific™ TSX Universal Series General Purpose Ultra-Low Freezers
Thermo Scientific™ TDE Series -86°C Ultra-Low Temperature Chest Freezers, General Purpose
Powered by H-drive's hydrocarbon technology, these systems save up to 25% in energy usage and heat rejection over conventional refrigerant ultra-low freezers.
Thermo Scientific™ TDE Series -86°C Ultra-Low Temperature Chest Freezers, General Purpose
Complete Your Order - Great Deals
Tempshield™ Cryo-Gloves™ Blue Waterproof Mid-Arm Length Cryogenic Gloves
A thin, seamless 100% waterproof liner provides protection from spills and splashes. State-of-the art materials and multi-layered construction provide cryogenic protection for ultra-cold applications down to -196°C (-320°F).
Tempshield™ Cryo-Gloves™ Blue Waterproof Mid-Arm Length Cryogenic Gloves
Cryogenic and Freezer Boxes
Cryogenic freezer boxes are containers designed to store biological samples at extremely low temperatures, typically below -150°C (-238°F) using liquid nitrogen or other cryogenic agents. These boxes are commonly used in laboratory settings to preserve the integrity and viability of sensitive samples, such as cells, tissues, and genetic materials.
Cryogenic and Freezer Boxes
Fisherbrand™ Permanent Markers
Fisherbrand™ Permanent Marking Pens write on glass, film, wood, plastic, metal, rubber, and porcelain. Perfect for marking sample bags, microscope or photographic slides, laboratory glassware and more. Write on all surfaces.
Fisherbrand™ Permanent Markers
FAQ
Ultra low temperature freezers are used to store sensitive biological and chemical samples at around -80°C. They support long-term sample integrity in research, pharmaceutical, clinical, and biobank laboratories using brands such as Thermo Scientific and Eppendorf.
Ultra low temperature freezers use advanced insulation, cascade refrigeration systems, and digital monitoring to maintain stable internal temperatures. Manufacturers like Binder and Thermo Scientific focus on uniformity and fast recovery after door openings.
Research labs, biobanks, pharmaceutical facilities, and clinical laboratories require ultra low temperature freezers to protect enzymes, vaccines, DNA, RNA, and other critical samples under controlled storage conditions.
Energy efficiency reduces operating costs and environmental impact while ensuring reliable storage. Modern ULT freezers from Thermo Scientific, Binder, and Fisherbrand are designed to balance performance with reduced power consumption.
Top Brands