Universal Pipette Tips
Universal pipette tips are compatible with pipettes from various manufacturers and have comparable performance to branded tips. Suitable for any genomic or biological application, these disposable tips are available in sterile and non-sterile versions for a variety of specific functions and in various styles and lengths.
Fisherbrand™ SureOne™ Aerosol Barrier Pipette Tips
Ensure accuracy and prevent contamination.
Contec™ NeutraKlean™ Detergent with Purified Water, Sterile
Eppendorf Quality™ epT.I.P.S™ Reloads
Reliable tips for precise pipetting.
Contec™ Non-Sterile CyChlor
Complete Your Order - Great Deals
Corning™ Costar™ Sterile Disposable Reagent Reservoirs
Manufactured from modified polystyrene for strength, Corning™ Costar™ Sterile Disposable Reagent Reservoirs are ideal for repetitive filling of multichannel pipettors..
Lorem Ipsum
Brand™ Polypropylene Stackable Tip Box
BRAND™ Polypropylene Stackable Tip Boxes are supplied with a tip tray. Filter and pipette tips are sold separately.
Lorem Ipsum
Daniels Healthcare™ Sharpsguard™ Polypropylene General Purpose Sharps Container
Use this general-purpose sharps container for outstanding protection against penetrating sharps and leaking fluids. Designed to meet demands for strength, simplicity, and value.
Lorem Ipsum
FAQ
Universal pipettes offer several benefits in a laboratory setting, including versatility in handling various types of liquids and applications. They are designed to be compatible with a wide range of tips and volumes, making them suitable for diverse experimental needs. Universal pipettes provide precision and accuracy in liquid handling, ensuring consistent and reliable sample dispensing. They are often ergonomically designed to reduce user fatigue and improve comfort during prolonged use. Additionally, universal pipettes are typically easy to calibrate and maintain, enhancing their longevity and performance in the lab.
Proper maintenance and calibration of universal pipettes is crucial for ensuring their accuracy and longevity. Regular cleaning with appropriate cleaning solutions prevents contamination and residue buildup. Periodic calibration according to the manufacturer's instructions or laboratory standards (such as ISO 8655) involves checking the accuracy and precision of the pipette using known volumes of liquid. Regular inspection for any signs of wear or damage, such as cracked seals or worn-out tips, is essential. Storing the pipette in an upright position when not in use prevents liquid from entering the internal components. Consider professional servicing and calibration services to ensure the pipette remains in optimal condition.
Common issues with universal pipettes include inconsistent volume dispensing, which can be caused by worn-out seals or improper calibration. Regular maintenance and calibration can help resolve this issue. Leaking can occur due to damaged or improperly seated tips, so ensure that tips are correctly attached and replace any damaged components. Jamming can result from not cleaning the pipette regularly; regular cleaning and proper handling can prevent this. User fatigue is another issue, as manual pipetting can cause hand fatigue over time. Using ergonomic pipettes and taking regular breaks can help reduce fatigue. By addressing these common issues through proper maintenance and handling, you can ensure the reliable performance of your universal pipettes.
Top Brands