Every Space In the Fume Hood - Frequently Asked Questions
Electronic single-channel pipettes feature electronic mechanisms that provide precise and accurate dispensing of liquids, minimizing human errors associated with manual pipetting. They often offer customizable settings, ergonomic design, and intuitive user interfaces for enhanced pipetting performance.
Regular maintenance of electronic pipettes includes calibrating according to manufacturer guidelines, ensuring proper charging or battery replacement, cleaning the pipette and accessories regularly, and storing them in a clean and dry environment when not in use.
Consider factors such as volume range, accuracy, precision, ergonomics, and compatibility with tips and accessories when selecting a manual single-channel pipette. Choose a pipette that suits your specific pipetting needs and workflow requirements.
Ensure proper technique by holding the pipette correctly, pre-wetting tips, dispensing at a consistent speed, and avoiding air bubbles in the tip. Regularly calibrate and maintain pipettes according to manufacturer guidelines to ensure accurate and reliable performance over time.
When selecting a lab coat, consider factors such as material (e.g., cotton, polyester, or a blend), size, durability, and additional features like pockets or knit cuffs. Choose a lab coat that provides comfort, mobility, and adequate protection for your specific laboratory tasks and safety requirements.