nginx
Every Space On the Shelf - Frequently Asked Questions
Universal pipette tips are designed to fit a wide range of pipettes from various manufacturers, offering compatibility with different pipette models and brands. Unlike regular pipette tips, which may be specific to certain pipette brands or models, universal tips provide versatility and convenience for laboratories with multiple types of pipettes.
When selecting universal pipette tips, consider factors such as volume range, material quality (e.g., polypropylene), sterility, and special features like filter tips or low-retention surfaces. Choose tips that ensure accurate and precise dispensing while minimizing sample waste and contamination.
Proper usage of Petri dishes involves sterilizing them before use, pouring agar medium into the dishes under aseptic conditions, and sealing them with lids to prevent contamination. Handle Petri dishes with clean hands or sterile gloves, avoid touching the agar surface, and incubate them in a suitable environment to promote microbial growth.
pH paper and test strips are simple, paper-based strips impregnated with pH-sensitive chemicals that change color in response to the acidity or alkalinity of a solution. They provide a quick and convenient method for measuring pH levels in various liquids, including water, urine, and saliva, by comparing the color change to a color chart.
Consider factors such as pH range, accuracy, sensitivity, ease of use, and compatibility with specific sample types when selecting pH paper and test strips. Choose strips that offer the appropriate pH range and resolution for your testing requirements, ensuring reliable and reproducible results.
When selecting centrifuge tubes, consider factors such as tube material, volume capacity, compatibility with centrifuge rotor types (e.g., fixed-angle or swinging bucket), and sealing properties (e.g., screw-cap or snap-cap). Choose tubes that are chemically resistant, autoclavable, and suitable for the specific centrifugation speeds and sample types required for your experiments.