Cell Culture Microplates
Cell culture microplates are essential consumables for in vitro cell growth, analysis, and screening across research, pharmaceutical, and biotechnology laboratories. Designed to support consistent cell attachment and proliferation, these plates are available in multiple well formats, surface treatments, and optical properties to meet diverse experimental needs. Discover reliable solutions from leading brands to support reproducible and scalable cell culture workflows.
Thermo Scientific™ Nunc™ MicroWell™ 96-Well, Nunclon Delta-Treated, Flat-Bottom Microplate
Optimized for robotics, automated readers and liquid handling systems, 96-well microplates are used for antibiotic screens, cell-based assays and screening compounds. The flat bottom shape is ideal for microscopic and optical measurements.
Thermo Scientific™ Nunc™ MicroWell™ 96-Well, Nunclon Delta-Treated, Flat-Bottom Microplate
Thermo Scientific™ Nunc™ Cell-Culture Treated Multidishes
Nunc Cell-Culture Treated Multidishes receive the proprietary Nunclon Delta surface treatment that promotes maximum adhesion for a broad range of cell types.
Thermo Scientific™ Nunc™ Cell-Culture Treated Multidishes
Thermo Scientific™ Nunc™ 96-Well Optical-Bottom Microplate, Black, TC Surface
Achieve optimal fluorescence and luminescence in cell culture and microscopic applications with Nunc MicroWell 384-well optical-bottom plates.
Thermo Scientific™ Nunc™ 96-Well Optical-Bottom Microplate, Black, TC Surface
Save Now - Exclusive Deals
Complete Your Order - Great Deals
Corning™ Costar™ Sterile Disposable Reagent Reservoirs
Manufactured from modified polystyrene for strength, Corning™ Costar™ sterile disposable reagent reservoirs are ideal for repetitive filling of multichannel pipettes.
Corning™ Costar™ Sterile Disposable Reagent Reservoirs
Pipette Specific Tips
Pipette specific tips are disposable and designed to fit specific manufacturers’ single and multichannel pipettes. Available sterile or non-sterile and in a range of sizes, styles, and colors, with and without filters.
Pipette Specific Tips
Manual Multichannel Pipettes
Manual multichannel pipettes are used to simultaneously transfer multiple batches of liquid between labware of different formats. Usually configured for 96-well or 384-well plates and for various volume ranges.
Manual Multichannel Pipettes
FAQ
Cell culture microplates are used to grow, maintain, and analyze cells in controlled in vitro environments. They support applications such as drug screening, toxicity testing, and cell-based assays using brands like Corning and Thermo Scientific Nunc.
The correct well format depends on throughput and assay volume. 96- and 384-well plates support high-throughput screening, while 6- and 24-well plates are ideal for routine cell culture and microscopy.
TC-treated and Nunclon Delta surfaces enhance cell attachment and growth. These treatments are commonly used for adherent cell lines and are offered by Corning, Thermo Scientific Nunc, and Fisherbrand.
Optical clarity enables accurate imaging and fluorescence or luminescence detection. Optical-bottom plates from Corning and Thermo Scientific are designed to improve signal quality in advanced assays.
All listed microplates have Accessibility = 15, meaning they are set up for use and availability across all supported European countries.
Top Brands