Centrifugal Filter Devices
Centrifugal filter devices help you concentrate and purify biological samples quickly using standard centrifuges. Choose the right MWCO and membrane chemistry to support protein concentration, buffer exchange, desalting, and nucleic-acid cleanup with consistent recovery. Explore high-performing options from Merck, Sartorius, Epredia, Cytiva, Corning, and Thermo Scientific—filtered for broad EU availability.
Useful Links
Save Now - Exclusive Deals
Product Code 10781543
Product Code 10310361
Complete Your Order - Great Deals
Must Have
Product Code 10188431
Product Code 10282244
Product Code 10088753
Product Code 10341782
Product Code 10704645
Product Code 10506282
FAQ
Centrifugal filter devices are used to concentrate and clean up samples by spinning liquid through a membrane. Common workflows include protein concentration, buffer exchange, desalting, and removing particulates before analysis. Popular options include Merck Amicon, Sartorius Vivaspin, and Corning Spin-X formats for routine lab centrifuges.
Choose an MWCO that retains your target while allowing smaller molecules to pass. A practical rule is selecting an MWCO 3–10× lower than the target protein’s molecular weight, then optimizing recovery. Merck and Sartorius ranges cover common MWCO steps to match concentration and buffer-exchange goals.
Membrane chemistry affects binding and compatibility. Low-binding membranes help preserve proteins and sensitive biomolecules, while other chemistries may suit clarification or nucleic-acid workflows. Review device specs and your buffer composition, then compare trusted lines from Merck, Sartorius, Cytiva, Corning, and Thermo Scientific for the best fit.
To reduce clogging, pre-clarify samples (spin down debris or pre-filter), avoid overloading viscous or particulate-rich solutions, and follow recommended spin speeds. Using the correct device size also helps maintain flow. Corning Spin-X style clarification devices and Merck/Sartorius concentrators support smoother downstream prep.
Low recovery can come from membrane binding, over-spinning to dryness, or selecting an MWCO that’s too high for the target. Use recommended maximum concentration factors, stop spins earlier, and rinse or recover from the retentate properly. Merck Amicon and Sartorius Vivaspin guidance can help standardize yields.
Use this merchandising page’s Featured Products, Deals table, and Brand Filters to focus on items set up broadly across the EU. You can also compare options by brand—Merck, Sartorius, Epredia, Cytiva, Corning, and Thermo Scientific—and then select the right size, MWCO, and membrane for your workflow.