Particulate Filtering Facepiece Respirators
Particulate Filtering Facepiece Respirators
Wearing proper respiratory protection is critical when you work in demanding environments such as the automotive industry, construction, the oil and gas industry, the chemical industry, public utilities, and many others. Disposable respirators protect workers against airborne particles in both oil and non-oil environments.
FEATURED
3M™ Particulate Respirator, 8300 Series
Comfortable, effective respiratory protection against dust particles and/or non-volatile liquid particles
3M™ Particulate Respirator, 8300 Series
GREAT VALUE
3M™ Particulate Respirators
Breathe easy with 3M™ Particulate Respirators
Reusable respirators with enhanced comfort, better fit, and great value.
3M™ Particulate Respirators
CONVEX SHAPE
3M™ Disposable Particulate Respirator, 8000 Series
Ensure comfortable wear over a range of face sizes
3M™ Disposable Particulate Respirator, 8000 Series
Selector Guide
Get the best-fit respiratory protection for your lab protocols with our interactive selection guide.
Save Now - Exclusive Deals
Complete Your Order - Great Deals
FAQ
Respirators and surgical masks are personal protective equipment (PPE) used by healthcare workers and non-healthcare workplaces. Respirators help reduce the wearer's exposure to airborne particles. Tight-fitting respirators, such as the N95, are designed to seal the wearer's face and are tested to meet minimum filtration performance requirements and other regulatory standards. A surgical mask prevents biological particles such as bacteria and viruses from being expelled from the wearer into the environment. Surgical masks are not necessarily designed to fit tightly against the face, allowing air to escape at the edges. Respirator are used in high-risk environments and areas with airborne infectious agents. Surgical mask are used in medical settings to protect against large respiratory droplets and prevent infection spread.
To correctly remove a filtering facepiece respirator:
  • Clean your hands
  • Avoid touching the front of the respirator, as it may be contaminated
  • Remove the bottom strap
  • Remove the top strap over your head, being careful not to let the respirator touch your face or clothes
  • Discard the respirator if it is a single-use type, or if reusable, place it in a clean, breathable container, like a paper bag, for future use. They should be stored indoors, in their original packaging, in a structured storage space where they can’t become crushed or distorted.
  • Clean your hands again immediately after removing the respirator.
N95 respirators are effective at filtering at least 95% of airborne particles, including:
  • Dust
  • Mist
  • Fumes
  • Smoke
  • Biological particles (e.g., bacteria and viruses)
  • Non-oil-based aerosols
N95 respirators are not resistant to oil, hence they are not suitable for filtering oil-based particles. They are commonly used in healthcare settings and various industries to protect against respiratory hazards.
Resources and Related Information
Three Step Guide to Selecting the Right Disposable Respirator
A guide to assist you in choosing the disposable respiratory product that best matches your application and comfort preferences.
Three Step Guide to Selecting the Right Disposable Respirator
PPE Selection, Fit & Maintenance Tools
Find information on product selection, fit, usage and maintenance for Personal Protective Equipment items.
PPE Selection, Fit & Maintenance Tools
Filtering Respirators
Learn about everything related to filtering respirators.
Filtering Respirators
Shop By Brand