Laboratory Freezers
Laboratory freezers are used to store a variety of samples between −10°C and −30°C, including pharmaceutical, biological, and other commonly used laboratory materials. Products may be upright, freestanding, or under-bench models providing a range of sizes and capacities. An automatic defrost option is available on some products.
Liebherr™ Spark-Free Upright Lab Freezer With Shelves
Secure, accurate and ultra energy-efficient, spark-free interior, freezer storage, the Liebherr™ Spark-Free Interior Lab Freezer is designed for general purpose applications with Liebherr build-quality you can rely on.
Contec™ Sterile IPA
Thermo Scientific™ TSG General Purpose Freezers - Boxed
The TSG Series of general purpose freezers are built with the performance, security, and quality features you have come to expect from Thermo Scientific™.
Contec™ Sterile IPA
Liebherr™ Perfection Upright Freezer for Scientific Use
Secure, accurate and ultra energy-efficient freezer storage of temperature-sensitive products, the Liebherr™ upright lab freezer is fully-equipped for lab applications with Liebherr™ build-quality you can rely on. 
Contec™ Sterile IPA
Complete Your Order - Great Deals
Cryogenic and Freezer Boxes
Cryogenic freezer boxes are containers designed to store biological samples at extremely low temperatures, typically below -150°C (-238°F) using liquid nitrogen or other cryogenic agents.
Lorem Ipsum
Storage Tubes
Intended for general laboratory storage, our storage tubes are available in various sizes and materials, and with various RCF ratings. Sterile or non-sterile, they may be suitable for autoclaving and may include barcode labels, tube caps, and/or racks. Some are suitable for freezer storage.
Lorem Ipsum
Labels and Sign Makers
Labels and sign makers are designed to meet all of your safety and facility needs including laboratory identification and traceability. They are available in portable or benchtop models with integrated labeling apps and/or software. 
Lorem Ipsum
FAQ
Regular maintenance is crucial for the optimal performance of -80°C freezers. It is recommended to perform routine checks every three to six months. This includes cleaning the filters, checking the door seals, and ensuring that the temperature control systems are functioning correctly. Annual professional servicing is also advised to address any potential issues and prolong the lifespan of the freezer.
Most -80°C freezers come with built-in battery backup systems that can maintain the temperature for a limited period during power outages. Additionally, many hospital labs invest in external backup generators that can provide power for extended periods. It is essential to regularly test these backup systems to ensure they are operational when needed.
The lifespan of a -80°C freezer can vary depending on the brand, model, and usage conditions. On average, these freezers can last between 10 to 15 years with proper maintenance. Regular servicing and adherence to manufacturer guidelines can help maximize the freezer's operational life, ensuring reliable performance over the years.
By understanding the critical role of ultra-low temperature storage and the key features of -80°C freezers, hospital labs can make informed decisions to enhance their sample preservation capabilities, ultimately supporting better patient outcomes and advancing medical research.
Related Documentation
Sample Storage Solutions
Thermo Scientific Storage Equipment and Consumables
PDF Image
Top Brands