Every Space On the Shelf
Stock your lab with a comprehensive selection of essentials and equipment to meet all your laboratory needs.
Stockroom Staples: Must Have Products to Keep Your Shelves Fully Stocked
From maintaining a well-stocked stockroom to organizing your shelves and cabinets, our range of products ensures that you are fully equipped for any project that comes your way.
Green Solutions
Greener Switch
Would you like to know what Greener Choice options are available for the laboratory products you buy?
FAQ
Universal pipette tips are designed to fit a wide range of pipettes from various manufacturers, offering compatibility with different pipette models and brands. Unlike regular pipette tips, which may be specific to certain pipette brands or models, universal tips provide versatility and convenience for laboratories with multiple types of pipettes.
Proper usage of Petri dishes involves sterilizing them before use, pouring agar medium into the dishes under aseptic conditions, and sealing them with lids to prevent contamination. Handle Petri dishes with clean hands or sterile gloves, avoid touching the agar surface, and incubate them in a suitable environment to promote microbial growth.
pH paper and test strips are simple, paper-based strips impregnated with pH-sensitive chemicals that change color in response to the acidity or alkalinity of a solution. They provide a quick and convenient method for measuring pH levels in various liquids, including water, urine, and saliva, by comparing the color change to a color chart.
When selecting centrifuge tubes, consider factors such as tube material, volume capacity, compatibility with centrifuge rotor types (e.g., fixed-angle or swinging bucket), and sealing properties (e.g., screw-cap or snap-cap). Choose tubes that are chemically resistant, autoclavable, and suitable for the specific centrifugation speeds and sample types required for your experiments.
Other Programs
Sustainability Program
Read more about how we can reach net-zero goals together.
Sustainability Program
Fisher Scientific Edge Program
Ensure quick dispatch of our most popular products.
Edge banner
Setting Up a New Lab?
Get all the support needed for setting up a new lab or expanding an existing lab through the NLSU program.
NLSU banner
News Corner
Explore the latest innovations within the Fisher Scientific Channel.
News Corner Program Image