Learn More
17alpha-Ethynyl-13C2-estradiol, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Ethynyl Estradiol-13C2 |
| CAS | 2483735-63-1 |
| Molecular Formula | 13C2 C18 H24 O2 |
| Quantity | 0.005 g |
| Synonym | 19-Norpregna-1,3,5(10)-trien-20-yne-3,17-diol-20,21-13C2, (17α)- (ACI), (17α)-19-Norpregna-1,3,5(10)-trien-20-yne-3,17-diol-20,21-13C2 (ACI), (17α)-19-Norpregna-1,3,5(10)-trien-20-yne-20,21-13C2-3,17-diol, Ethynyl-13C2 estradiol, Ethinylestradiol (ethinyl-13C2) |
| InChI Formula | InChI=1 S/C20H24O2/c1-3-20(22)11-9-18-17-6-4-13-12-14(21)5-7-15(13)16(17)8-10-19(18,20)2/h1,5,7,12,16-18,21-22 H,4,6,8-11H2,2H3/t16-,17-,18+,19+,20+/m1/s1/i1+1,3+1 |
| SMILES | C[C@]12 CC[C@H]3 [C@@H](CCc4cc(O)ccc34)[C@@H]1 CC[C@@]2(O)[13 C]#[13 CH] |
| IUPAC Name | (8 R,9 S,13 S,14 S,17 S)-17-(1,2-^{13}C_{2})ethynyl-13-methyl-7,8,9,11,12,14,15,16-octahydro-6 H-cyclopenta[a]phenanthrene-3,17-diol |
| Molecular Weight (g/mol) | 298.3887 |
| Formula Weight | 298.1843 g/mol |
| Show More |