missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-2-(2,4-Dichlorophenoxy-d3)propionic Acid, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Dichlorprop-D3 (phenoxy-D3) |
| CAS | 1219803-66-3 |
| Molecular Formula | C9 D3 H5 Cl2 O3 |
| Quantity | 0.01 g |
| Synonym | 2-(2,4-Dichlorophenoxy-3,5,6-d3)propanoic acid, (±)-2-(2,4-Dichlorophenoxy-d3)propionic Acid, Dichlorprop-d3, Dichlorprop-D3 (phenoxy-D3) |
| InChI Formula | InChI=1 S/C9H8Cl2O3/c1-5(9(12)13)14-8-3-2-6(10)4-7(8)11/h2-5 H,1H3,(H,12,13)/i2D,3 D,4 D |
| SMILES | [2 H]c1c([2 H])c(OC(C)C(=O)O)c(Cl)c([2 H])c1Cl |
| IUPAC Name | 2-(2,4-dichloro-3,5,6-trideuteriophenoxy)propanoic acid |
| Molecular Weight (g/mol) | 238.0825 |
| Formula Weight | 237.0039 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction