Learn More
2,4',5-Trichlorobiphenyl-2',3',5',6'-d4, CDN
Click to view available options
Quantity:
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | 2,4',5-Trichlorobiphenyl-2',3',5',6'-D4 |
| CAS | 1219794-87-2 |
| Molecular Formula | C12 D4 H3 Cl3 |
| Quantity | 0.01 g |
| Synonym | 2,4',5-Trichlorobiphenyl-2',3',5',6'-d4, 2,4',5-Trichloro-1,1'-biphenyl-2',3',5',6'-d4, 2,4',5-Trichlorobiphenyl-2',3',5',6'-D4, 4,2',5'-Trichlorobiphenyl-2,3,5,6-d4, PCB 31 D4, 2,4',5-TriCB-2',3',5',6'-d4 |
| InChI Formula | InChI=1 S/C12H7Cl3/c13-9-3-1-8(2-4-9)11-7-10(14)5-6-12(11)15/h1-7 H/i1D,2 D,3 D,4 D |
| SMILES | [2 H]c1c([2 H])c(c([2 H])c([2 H])c1Cl)c2cc(Cl)ccc2Cl |
| IUPAC Name | 1-chloro-2,3,5,6-tetradeuterio-4-(2,5-dichlorophenyl)benzene |
| Molecular Weight (g/mol) | 261.5676 |
| Formula Weight | 259.9864 g/mol |
| Show More |