missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Estren-17alpha-methyl-d3-17beta-ol-3-one, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Normethandrone-20,20,20-D3 |
| Molecular Formula | C19 D3 H25 O2 |
| Quantity | 0.01 g |
| Synonym | Normethandrone-d3, Normethandrone-20,20,20-D3, (17β)-17-Hydroxy-17-(methyl-d3)estr-4-en-3-one, 17-(Methyl-d3)-19-nortestosterone |
| InChI Formula | InChI=1 S/C19H28O2/c1-18-9-7-15-14-6-4-13(20)11-12(14)3-5-16(15)17(18)8-10-19(18,2)21/h11,14-17,21 H,3-10H2,1-2H3/t14-,15+,16+,17-,18-,19-/m0/s1/i2D3 |
| SMILES | [2 H]C([2 H])([2 H])[C@]1(O)CC[C@H]2 [C@@H]3 CCC4=CC(=O)CC[C@@H]4 [C@H]3 CC[C@]12 C |
| IUPAC Name | (8 R,9 S,10 R,13 S,14 S,17 S)-17-hydroxy-13-methyl-17-(trideuteriomethyl)-1,2,6,7,8,9,10,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthren-3-one |
| Molecular Weight (g/mol) | 291.4429 |
| Formula Weight | 291.2278 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction